Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133429829-133430364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS13
Entrez Gene ID
11093
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 13
Gene Aliases
ADAM-TS13, ADAMTS-13, C9orf8, VWFCP, vWF-CP
UniGene
Hs.131433
Gene Chromosome Location
Chr.9: 133414339-133459403 on Build GRCh38
Transcripts
NM_139025.4 NM_139026.4 NM_139027.4 NR_024514.2 XM_011518176.2 XM_017014232.1 XM_017014233.1 XM_017014234.1 XM_017014235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847971 c.986C>T p.S329L Chr.9: 133429861-133429861 on Build GRCh38
5731664 c.945C>T p.F315F Chr.9: 133430059-133430059 on Build GRCh38
6319829 c.844G>T p.V282L Chr.9: 133429958-133429958 on Build GRCh38
6390931 c.977G>A p.R326K Chr.9: 133430091-133430091 on Build GRCh38
1106434 c.920C>T p.A307V Chr.9: 133430034-133430034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587673969 Chr.9: 133429892-133429892 on Build GRCh38
rs782114348 Chr.9: 133430005-133430005 on Build GRCh38
rs1015482894 Chr.9: 133430209-133430209 on Build GRCh38
rs587658314 Chr.9: 133430255-133430255 on Build GRCh38
rs782068918 Chr.9: 133430196-133430196 on Build GRCh38
rs587703137 Chr.9: 133430236-133430236 on Build GRCh38
rs372976387 Chr.9: 133429881-133429881 on Build GRCh38
rs782402561 Chr.9: 133430148-133430148 on Build GRCh38
rs781911708 Chr.9: 133430063-133430063 on Build GRCh38
rs782268976 Chr.9: 133430086-133430086 on Build GRCh38
rs970049597 Chr.9: 133429925-133429925 on Build GRCh38
rs946491640 Chr.9: 133430186-133430186 on Build GRCh38
rs377143536 Chr.9: 133430017-133430017 on Build GRCh38
rs782579493 Chr.9: 133430162-133430162 on Build GRCh38
rs902169055 Chr.9: 133430056-133430056 on Build GRCh38
rs1028751124 Chr.9: 133430263-133430263 on Build GRCh38
rs782022887 Chr.9: 133429862-133429862 on Build GRCh38
rs782542948 Chr.9: 133430068-133430068 on Build GRCh38
rs34254995 Chr.9: 133429934-133429934 on Build GRCh38
rs782360062 Chr.9: 133430082-133430082 on Build GRCh38
rs1007140315 Chr.9: 133430095-133430095 on Build GRCh38
rs587744487 Chr.9: 133430167-133430167 on Build GRCh38
rs929575982 Chr.9: 133429932-133429932 on Build GRCh38
rs782170431 Chr.9: 133429907-133429917 on Build GRCh38
rs587615316 Chr.9: 133429883-133429883 on Build GRCh38
rs782093351 Chr.9: 133430123-133430124 on Build GRCh38
rs782031676 Chr.9: 133430108-133430108 on Build GRCh38
rs903657184 Chr.9: 133429875-133429875 on Build GRCh38
rs782608274 Chr.9: 133430304-133430304 on Build GRCh38
rs587600662 Chr.9: 133429913-133429913 on Build GRCh38
rs753339966 Chr.9: 133429888-133429888 on Build GRCh38
rs36219895 Chr.9: 133430338-133430338 on Build GRCh38
rs782744649 Chr.9: 133430164-133430165 on Build GRCh38
rs28729234 Chr.9: 133430112-133430112 on Build GRCh38
rs995701451 Chr.9: 133429965-133429965 on Build GRCh38
rs1029520931 Chr.9: 133429876-133429876 on Build GRCh38
rs782059317 Chr.9: 133430023-133430023 on Build GRCh38
rs782640937 Chr.9: 133430059-133430059 on Build GRCh38
rs1053205257 Chr.9: 133429882-133429882 on Build GRCh38
rs782254917 Chr.9: 133430175-133430175 on Build GRCh38
rs782183226 Chr.9: 133430163-133430163 on Build GRCh38
rs782102945 Chr.9: 133430001-133430001 on Build GRCh38
rs782429486 Chr.9: 133429979-133429979 on Build GRCh38
rs781870655 Chr.9: 133430127-133430127 on Build GRCh38
rs782067565 Chr.9: 133429867-133429867 on Build GRCh38
rs375767052 Chr.9: 133430134-133430134 on Build GRCh38
rs781828222 Chr.9: 133430145-133430145 on Build GRCh38
rs587671802 Chr.9: 133429948-133429948 on Build GRCh38
rs367819139 Chr.9: 133430147-133430147 on Build GRCh38
rs150518374 Chr.9: 133430033-133430033 on Build GRCh38
rs782291570 Chr.9: 133430055-133430055 on Build GRCh38
rs1043474009 Chr.9: 133430187-133430187 on Build GRCh38
rs966252312 Chr.9: 133430245-133430245 on Build GRCh38
rs372413496 Chr.9: 133430002-133430002 on Build GRCh38
rs587667471 Chr.9: 133430087-133430087 on Build GRCh38
rs782701360 Chr.9: 133429878-133429878 on Build GRCh38
rs931645668 Chr.9: 133430078-133430078 on Build GRCh38
rs782184693 Chr.9: 133429971-133429971 on Build GRCh38
rs367899892 Chr.9: 133429921-133429921 on Build GRCh38
rs369514834 Chr.9: 133430047-133430047 on Build GRCh38
rs36219562 Chr.9: 133430050-133430050 on Build GRCh38
rs281875336 Chr.9: 133430046-133430046 on Build GRCh38
rs782616591 Chr.9: 133430144-133430144 on Build GRCh38
rs1024280917 Chr.9: 133429906-133429906 on Build GRCh38
rs782282073 Chr.9: 133429988-133429988 on Build GRCh38
rs782276854 Chr.9: 133429894-133429894 on Build GRCh38
rs782740878 Chr.9: 133430126-133430126 on Build GRCh38
rs983444953 Chr.9: 133430154-133430154 on Build GRCh38
rs185197533 Chr.9: 133430237-133430237 on Build GRCh38
rs782674608 Chr.9: 133429958-133429958 on Build GRCh38
rs754541465 Chr.9: 133429984-133429984 on Build GRCh38
rs782140908 Chr.9: 133430296-133430296 on Build GRCh38
rs955285640 Chr.9: 133430065-133430065 on Build GRCh38
rs281875285 Chr.9: 133430025-133430025 on Build GRCh38
rs782322129 Chr.9: 133429989-133429989 on Build GRCh38
rs781808050 Chr.9: 133430141-133430141 on Build GRCh38
rs3124773 Chr.9: 133430170-133430170 on Build GRCh38
rs782420537 Chr.9: 133430178-133430178 on Build GRCh38
rs925233142 Chr.9: 133430115-133430115 on Build GRCh38
rs782001479 Chr.9: 133430004-133430004 on Build GRCh38
rs781863038 Chr.9: 133430170-133430170 on Build GRCh38
rs781918765 Chr.9: 133430114-133430114 on Build GRCh38
rs782601193 Chr.9: 133430075-133430075 on Build GRCh38
rs934998373 Chr.9: 133430224-133430224 on Build GRCh38
rs1050746608 Chr.9: 133430230-133430230 on Build GRCh38
rs587716091 Chr.9: 133429903-133429903 on Build GRCh38
rs782818563 Chr.9: 133430016-133430016 on Build GRCh38
rs782020311 Chr.9: 133430179-133430179 on Build GRCh38
rs1027855473 Chr.9: 133430173-133430173 on Build GRCh38
rs988091901 Chr.9: 133430343-133430343 on Build GRCh38
rs975797629 Chr.9: 133430257-133430257 on Build GRCh38
rs782613885 Chr.9: 133429982-133429982 on Build GRCh38
rs782798456 Chr.9: 133430136-133430136 on Build GRCh38
rs782444847 Chr.9: 133430142-133430142 on Build GRCh38
rs782142895 Chr.9: 133430111-133430111 on Build GRCh38
rs371350265 Chr.9: 133430153-133430153 on Build GRCh38
rs782379766 Chr.9: 133429986-133429986 on Build GRCh38
rs782325378 Chr.9: 133429906-133429907 on Build GRCh38
rs782517734 Chr.9: 133430146-133430146 on Build GRCh38
rs782596096 Chr.9: 133430083-133430083 on Build GRCh38
rs781919209 Chr.9: 133430062-133430062 on Build GRCh38
rs782799358 Chr.9: 133430340-133430340 on Build GRCh38
rs1048430091 Chr.9: 133429947-133429947 on Build GRCh38
rs1027868756 Chr.9: 133430010-133430010 on Build GRCh38
rs902241505 Chr.9: 133430223-133430223 on Build GRCh38
rs1030005935 Chr.9: 133430064-133430064 on Build GRCh38
rs935266108 Chr.9: 133430342-133430342 on Build GRCh38
rs781919201 Chr.9: 133429991-133429991 on Build GRCh38
rs782036565 Chr.9: 133429987-133429987 on Build GRCh38
rs587616629 Chr.9: 133430130-133430130 on Build GRCh38

View Full Product Details