Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133536383-133536881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL2
Entrez Gene ID
9719
Gene Name
ADAMTS like 2
Gene Aliases
GPHYSD1
UniGene
Hs.522543
Gene Chromosome Location
Chr.9: 133532164-133575519 on Build GRCh38
Transcripts
NM_001145320.1 NM_014694.3 XM_005272237.3 XM_005272238.3 XM_005272239.2 XM_011519241.2 XM_011519242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4511487 c.86C>T p.S29F Chr.9: 133536798-133536798 on Build GRCh38
5998513 c.46G>C p.V16L Chr.9: 133536758-133536758 on Build GRCh38
5998514 c.373G>C p.V125L Chr.9: 133536758-133536758 on Build GRCh38
5668895 c.392G>T p.G131V Chr.9: 133536777-133536777 on Build GRCh38
6333977 c.52G>T p.A18S Chr.9: 133536764-133536764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747085893 Chr.9: 133536570-133536570 on Build GRCh38
rs141076725 Chr.9: 133536466-133536466 on Build GRCh38
rs970692815 Chr.9: 133536518-133536518 on Build GRCh38
rs761336043 Chr.9: 133536457-133536457 on Build GRCh38
rs761794484 Chr.9: 133536594-133536594 on Build GRCh38
rs756148714 Chr.9: 133536745-133536745 on Build GRCh38
rs150194394 Chr.9: 133536844-133536844 on Build GRCh38
rs552420818 Chr.9: 133536545-133536545 on Build GRCh38
rs747347111 Chr.9: 133536854-133536854 on Build GRCh38
rs1041933816 Chr.9: 133536633-133536633 on Build GRCh38
rs765685064 Chr.9: 133536626-133536626 on Build GRCh38
rs868071341 Chr.9: 133536779-133536779 on Build GRCh38
rs62620179 Chr.9: 133536705-133536705 on Build GRCh38
rs773066046 Chr.9: 133536608-133536608 on Build GRCh38
rs965701356 Chr.9: 133536643-133536643 on Build GRCh38
rs375130193 Chr.9: 133536496-133536496 on Build GRCh38
rs983562673 Chr.9: 133536455-133536455 on Build GRCh38
rs924855974 Chr.9: 133536684-133536684 on Build GRCh38
rs773151671 Chr.9: 133536681-133536681 on Build GRCh38
rs779798976 Chr.9: 133536490-133536490 on Build GRCh38
rs762657949 Chr.9: 133536801-133536801 on Build GRCh38
rs148178122 Chr.9: 133536463-133536463 on Build GRCh38
rs769892909 Chr.9: 133536658-133536658 on Build GRCh38
rs1000264488 Chr.9: 133536521-133536521 on Build GRCh38
rs745375757 Chr.9: 133536794-133536794 on Build GRCh38
rs759076801 Chr.9: 133536622-133536622 on Build GRCh38
rs753539826 Chr.9: 133536581-133536581 on Build GRCh38
rs796441178 Chr.9: 133536456-133536456 on Build GRCh38
rs752122891 Chr.9: 133536443-133536443 on Build GRCh38
rs1007607833 Chr.9: 133536464-133536464 on Build GRCh38
rs746029004 Chr.9: 133536580-133536580 on Build GRCh38
rs199645279 Chr.9: 133536820-133536820 on Build GRCh38
rs778765212 Chr.9: 133536673-133536673 on Build GRCh38
rs759044523 Chr.9: 133536629-133536629 on Build GRCh38
rs771990994 Chr.9: 133536599-133536599 on Build GRCh38
rs904642378 Chr.9: 133536509-133536509 on Build GRCh38
rs757436707 Chr.9: 133536640-133536640 on Build GRCh38
rs781093264 Chr.9: 133536648-133536648 on Build GRCh38
rs533336549 Chr.9: 133536572-133536572 on Build GRCh38
rs11507717 Chr.9: 133536484-133536484 on Build GRCh38
rs756165826 Chr.9: 133536842-133536842 on Build GRCh38
rs1043530943 Chr.9: 133536507-133536507 on Build GRCh38
rs759293340 Chr.9: 133536688-133536688 on Build GRCh38
rs776273406 Chr.9: 133536793-133536793 on Build GRCh38
rs774836985 Chr.9: 133536809-133536809 on Build GRCh38
rs780264406 Chr.9: 133536751-133536751 on Build GRCh38
rs1022037527 Chr.9: 133536840-133536840 on Build GRCh38
rs970222661 Chr.9: 133536559-133536559 on Build GRCh38
rs930403470 Chr.9: 133536435-133536435 on Build GRCh38
rs770758713 Chr.9: 133536789-133536789 on Build GRCh38
rs374862671 Chr.9: 133536845-133536845 on Build GRCh38
rs764597370 Chr.9: 133536802-133536802 on Build GRCh38
rs781383669 Chr.9: 133536736-133536736 on Build GRCh38
rs775979762 Chr.9: 133536686-133536686 on Build GRCh38
rs372811359 Chr.9: 133536856-133536856 on Build GRCh38
rs765754783 Chr.9: 133536616-133536616 on Build GRCh38
rs755388728 Chr.9: 133536541-133536541 on Build GRCh38
rs754791574 Chr.9: 133536646-133536646 on Build GRCh38
rs760565627 Chr.9: 133536682-133536682 on Build GRCh38
rs773987435 Chr.9: 133536587-133536587 on Build GRCh38
rs369288950 Chr.9: 133536467-133536467 on Build GRCh38
rs371639887 Chr.9: 133536835-133536835 on Build GRCh38
rs769290624 Chr.9: 133536796-133536796 on Build GRCh38
rs758954520 Chr.9: 133536546-133536546 on Build GRCh38
rs199857142 Chr.9: 133536566-133536566 on Build GRCh38
rs752055478 Chr.9: 133536635-133536635 on Build GRCh38
rs549765000 Chr.9: 133536674-133536674 on Build GRCh38
rs537818418 Chr.9: 133536630-133536630 on Build GRCh38
rs145897018 Chr.9: 133536770-133536770 on Build GRCh38
rs757677138 Chr.9: 133536730-133536730 on Build GRCh38
rs920309644 Chr.9: 133536417-133536417 on Build GRCh38
rs760191683 Chr.9: 133536614-133536614 on Build GRCh38
rs369206306 Chr.9: 133536540-133536540 on Build GRCh38
rs750462029 Chr.9: 133536738-133536738 on Build GRCh38
rs758219925 Chr.9: 133536783-133536783 on Build GRCh38
rs989229970 Chr.9: 133536552-133536552 on Build GRCh38
rs62637569 Chr.9: 133536800-133536800 on Build GRCh38
rs762446501 Chr.9: 133536813-133536813 on Build GRCh38
rs1026174994 Chr.9: 133536488-133536488 on Build GRCh38
rs369787577 Chr.9: 133536510-133536510 on Build GRCh38
rs746144206 Chr.9: 133536651-133536651 on Build GRCh38
rs563146559 Chr.9: 133536458-133536458 on Build GRCh38
rs200378599 Chr.9: 133536723-133536723 on Build GRCh38
rs753555772 Chr.9: 133536621-133536621 on Build GRCh38
rs771796545 Chr.9: 133536678-133536678 on Build GRCh38
rs752331570 Chr.9: 133536709-133536709 on Build GRCh38
rs951910815 Chr.9: 133536604-133536604 on Build GRCh38
rs777512904 Chr.9: 133536785-133536785 on Build GRCh38
rs763424274 Chr.9: 133536717-133536717 on Build GRCh38
rs371158728 Chr.9: 133536683-133536683 on Build GRCh38
rs574275461 Chr.9: 133536539-133536539 on Build GRCh38
rs779104663 Chr.9: 133536544-133536544 on Build GRCh38
rs781491540 Chr.9: 133536574-133536574 on Build GRCh38

View Full Product Details