Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRP36
Entrez Gene ID
88745
Gene Name
ribosomal RNA processing 36
Gene Aliases
C6orf153, dJ20C7.4
Chromosome Location
Chr.6: 43021647 - 43029599 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033112.2 NP_149103.1 2 - 3 286 70
GenBank mRNA AK313816.1 - 2 - 3 296 70
BC006293.2 - 2 - 3 280 70
BC011933.2 - 2 - 3 286 70

View Full Product Details