The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012361.3 | NP_001012361.1 | 5 - 6 | 629 | 131 |
NM_145241.4 | NP_660284.1 | 5 - 6 | 626 | 131 | |
NR_134677.1 | 5 - 6 | 558 | 131 | ||
GenBank mRNA | AK093240.1 | - | 5 - 6 | 558 | 131 |
AK313843.1 | - | 5 - 6 | 568 | 131 | |
AL833635.1 | - | 5 - 6 | 475 | 131 | |
BC012352.1 | - | 5 - 6 | 558 | 131 | |
BC022982.1 | - | 5 - 6 | 555 | 131 |