The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031711.2 | NP_001026881.1 | 3 - 4 | 348 | 68 |
XM_011534597.1 | XP_011532899.1 | 3 - 4 | 241 | 68 | |
XM_011534598.2 | XP_011532900.1 | 3 - 4 | 322 | 68 | |
XM_017009654.1 | XP_016865143.1 | 3 - 4 | 470 | 68 | |
XM_017009655.1 | XP_016865144.1 | 3 - 4 | 463 | 68 | |
GenBank mRNA | AB033007.1 | - | 3 - 4 | 293 | 68 |
AF267855.1 | - | 1 - 2 | 249 | 68 | |
BC012766.2 | - | 3 - 4 | 230 | 68 | |
BC014490.1 | - | 3 - 4 | 223 | 68 |