Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
Chromosome Location
Chr.16: 57186284 - 57240475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305163.1 NP_001292092.1 7 - 8 1694 100
NM_001305164.1 NP_001292093.1 7 - 8 1073 100
NM_133368.2 NP_588609.1 7 - 8 1095 100
XM_005256220.1 XP_005256277.1 7 - 8 1402 100
XM_011523427.1 XP_011521729.1 8 - 9 1181 100
XM_011523428.1 XP_011521730.1 8 - 9 1495 100
XM_011523430.1 XP_011521732.1 8 - 9 1122 100
XM_017023844.1 XP_016879333.1 8 - 9 1868 100
GenBank mRNA AB075852.1 - 7 - 8 1015 100
AK027549.1 - 7 - 8 1645 100
AK172845.1 - 7 - 8 1051 100
BC013173.2 - 7 - 8 1050 100

View Full Product Details