Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM15
Entrez Gene ID
643155
Gene Name
small integral membrane protein 15
Gene Aliases
C5orf43
Chromosome Location
Chr.5: 61157709 - 61162475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048249.3 NP_001041714.1 2 - 3 403 91
GenBank mRNA AK291180.1 - 3 - 4 312 91
BC014242.2 - 2 - 3 257 91
BC047489.2 - 2 - 3 284 91
BC066550.1 - 2 - 3 382 91
BC103711.1 - 2 - 3 358 91
BC146876.1 - 2 - 3 248 91
BC146881.1 - 2 - 3 248 91
BX538015.1 - 2 - 3 398 91
BX648459.1 - 2 - 3 375 91
DA039277.1 - 2 - 3 403 91

View Full Product Details