Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137180371-137180867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC2
Entrez Gene ID
29882
Gene Name
anaphase promoting complex subunit 2
Gene Aliases
APC2
UniGene
Hs.533262
Gene Chromosome Location
Chr.9: 137174784-137188605 on Build GRCh38
Transcripts
NM_013366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599533 c.1597T>C p.F533L Chr.9: 137180801-137180801 on Build GRCh38
1701977 c.1603_1604CC>TT p.P535F Chr.9: 137180794-137180795 on Build GRCh38
4661482 c.1602C>T p.S534S Chr.9: 137180796-137180796 on Build GRCh38
6599535 c.1624G>A p.V542M Chr.9: 137180514-137180514 on Build GRCh38
1283403 c.1692G>C p.M564I Chr.9: 137180379-137180379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777359521 Chr.9: 137180397-137180397 on Build GRCh38
rs775355974 Chr.9: 137180412-137180412 on Build GRCh38
rs769745791 Chr.9: 137180821-137180821 on Build GRCh38
rs779533020 Chr.9: 137180543-137180543 on Build GRCh38
rs746457748 Chr.9: 137180398-137180398 on Build GRCh38
rs888088466 Chr.9: 137180599-137180599 on Build GRCh38
rs914207111 Chr.9: 137180667-137180667 on Build GRCh38
rs762124027 Chr.9: 137180836-137180836 on Build GRCh38
rs370157791 Chr.9: 137180422-137180422 on Build GRCh38
rs534830248 Chr.9: 137180676-137180676 on Build GRCh38
rs563493064 Chr.9: 137180760-137180760 on Build GRCh38
rs1057220524 Chr.9: 137180604-137180604 on Build GRCh38
rs762319899 Chr.9: 137180754-137180754 on Build GRCh38
rs370349824 Chr.9: 137180413-137180413 on Build GRCh38
rs753429595 Chr.9: 137180778-137180778 on Build GRCh38
rs928526195 Chr.9: 137180686-137180686 on Build GRCh38
rs766756289 Chr.9: 137180419-137180419 on Build GRCh38
rs959944538 Chr.9: 137180656-137180656 on Build GRCh38
rs900620771 Chr.9: 137180727-137180727 on Build GRCh38
rs912188448 Chr.9: 137180758-137180758 on Build GRCh38
rs758445484 Chr.9: 137180799-137180799 on Build GRCh38
rs774718940 Chr.9: 137180835-137180835 on Build GRCh38
rs578138904 Chr.9: 137180711-137180711 on Build GRCh38
rs1034229846 Chr.9: 137180652-137180652 on Build GRCh38
rs777825594 Chr.9: 137180813-137180813 on Build GRCh38
rs368099052 Chr.9: 137180406-137180406 on Build GRCh38
rs200270965 Chr.9: 137180473-137180473 on Build GRCh38
rs189180559 Chr.9: 137180420-137180420 on Build GRCh38
rs28526257 Chr.9: 137180645-137180645 on Build GRCh38
rs368338340 Chr.9: 137180558-137180558 on Build GRCh38
rs570580805 Chr.9: 137180514-137180514 on Build GRCh38
rs200816137 Chr.9: 137180771-137180771 on Build GRCh38
rs762609231 Chr.9: 137180417-137180417 on Build GRCh38
rs111281027 Chr.9: 137180505-137180505 on Build GRCh38
rs989859078 Chr.9: 137180598-137180598 on Build GRCh38
rs755964405 Chr.9: 137180541-137180544 on Build GRCh38
rs938611182 Chr.9: 137180695-137180695 on Build GRCh38
rs1047568896 Chr.9: 137180703-137180703 on Build GRCh38
rs965075693 Chr.9: 137180470-137180470 on Build GRCh38
rs555810109 Chr.9: 137180647-137180647 on Build GRCh38
rs763429023 Chr.9: 137180750-137180750 on Build GRCh38
rs370825540 Chr.9: 137180421-137180421 on Build GRCh38
rs112208172 Chr.9: 137180538-137180538 on Build GRCh38
rs192899809 Chr.9: 137180457-137180457 on Build GRCh38
rs572371940 Chr.9: 137180669-137180669 on Build GRCh38
rs368628204 Chr.9: 137180436-137180436 on Build GRCh38
rs771131725 Chr.9: 137180478-137180478 on Build GRCh38
rs199926183 Chr.9: 137180793-137180793 on Build GRCh38
rs770598543 Chr.9: 137180399-137180399 on Build GRCh38
rs760807828 Chr.9: 137180418-137180418 on Build GRCh38
rs141269608 Chr.9: 137180819-137180819 on Build GRCh38
rs775846010 Chr.9: 137180825-137180825 on Build GRCh38
rs148237012 Chr.9: 137180788-137180788 on Build GRCh38
rs374238783 Chr.9: 137180426-137180426 on Build GRCh38
rs199697066 Chr.9: 137180515-137180515 on Build GRCh38
rs776788407 Chr.9: 137180483-137180483 on Build GRCh38
rs776876751 Chr.9: 137180566-137180566 on Build GRCh38
rs761218948 Chr.9: 137180560-137180560 on Build GRCh38
rs763192021 Chr.9: 137180521-137180521 on Build GRCh38
rs993906202 Chr.9: 137180576-137180576 on Build GRCh38
rs547784463 Chr.9: 137180432-137180432 on Build GRCh38
rs1031089066 Chr.9: 137180761-137180761 on Build GRCh38
rs373543166 Chr.9: 137180537-137180537 on Build GRCh38
rs745905905 Chr.9: 137180579-137180579 on Build GRCh38
rs775548357 Chr.9: 137180749-137180749 on Build GRCh38
rs978844795 Chr.9: 137180725-137180725 on Build GRCh38
rs780787610 Chr.9: 137180400-137180400 on Build GRCh38
rs575531529 Chr.9: 137180774-137180774 on Build GRCh38
rs569964436 Chr.9: 137180536-137180536 on Build GRCh38
rs748103801 Chr.9: 137180557-137180557 on Build GRCh38
rs745513587 Chr.9: 137180433-137180433 on Build GRCh38
rs779911105 Chr.9: 137180437-137180437 on Build GRCh38
rs143202554 Chr.9: 137180488-137180488 on Build GRCh38
rs754716539 Chr.9: 137180549-137180549 on Build GRCh38
rs550142312 Chr.9: 137180524-137180524 on Build GRCh38
rs371130727 Chr.9: 137180707-137180707 on Build GRCh38
rs770099036 Chr.9: 137180740-137180740 on Build GRCh38
rs749158011 Chr.9: 137180546-137180546 on Build GRCh38
rs745709786 Chr.9: 137180405-137180405 on Build GRCh38
rs768420187 Chr.9: 137180416-137180416 on Build GRCh38
rs371997907 Chr.9: 137180751-137180751 on Build GRCh38
rs768232660 Chr.9: 137180425-137180426 on Build GRCh38
rs374387269 Chr.9: 137180417-137180417 on Build GRCh38
rs778976859 Chr.9: 137180553-137180553 on Build GRCh38
rs1016245126 Chr.9: 137180440-137180440 on Build GRCh38
rs748986518 Chr.9: 137180443-137180443 on Build GRCh38
rs749473946 Chr.9: 137180826-137180826 on Build GRCh38
rs1030359926 Chr.9: 137180737-137180737 on Build GRCh38
rs76321625 Chr.9: 137180842-137180842 on Build GRCh38
rs771080792 Chr.9: 137180773-137180773 on Build GRCh38
rs531251486 Chr.9: 137180431-137180431 on Build GRCh38
rs955766356 Chr.9: 137180563-137180563 on Build GRCh38
rs73571568 Chr.9: 137180729-137180729 on Build GRCh38
rs764707481 Chr.9: 137180535-137180535 on Build GRCh38
rs758971802 Chr.9: 137180425-137180425 on Build GRCh38
rs1006271664 Chr.9: 137180710-137180710 on Build GRCh38
rs149530100 Chr.9: 137180464-137180464 on Build GRCh38
rs774628994 Chr.9: 137180752-137180752 on Build GRCh38

View Full Product Details