Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137181575-137182066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC2
Entrez Gene ID
29882
Gene Name
anaphase promoting complex subunit 2
Gene Aliases
APC2
UniGene
Hs.533262
Gene Chromosome Location
Chr.9: 137174784-137188605 on Build GRCh38
Transcripts
NM_013366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701978 c.1349G>A p.G450E Chr.9: 137181800-137181800 on Build GRCh38
1107058 c.1324C>T p.L442L Chr.9: 137181825-137181825 on Build GRCh38
1107057 c.1342G>A p.G448R Chr.9: 137181807-137181807 on Build GRCh38
3779858 c.1405G>A p.D469N Chr.9: 137181744-137181744 on Build GRCh38
1107056 c.1461C>T p.A487A Chr.9: 137181688-137181688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776533904 Chr.9: 137181897-137181897 on Build GRCh38
rs748770186 Chr.9: 137181776-137181776 on Build GRCh38
rs767544340 Chr.9: 137181639-137181640 on Build GRCh38
rs1028906022 Chr.9: 137181603-137181603 on Build GRCh38
rs374011505 Chr.9: 137181639-137181639 on Build GRCh38
rs201323692 Chr.9: 137181749-137181749 on Build GRCh38
rs1038964012 Chr.9: 137181920-137181920 on Build GRCh38
rs994945202 Chr.9: 137181599-137181599 on Build GRCh38
rs758447330 Chr.9: 137181821-137181821 on Build GRCh38
rs571333032 Chr.9: 137181936-137181936 on Build GRCh38
rs1046591288 Chr.9: 137182040-137182040 on Build GRCh38
rs752723453 Chr.9: 137181748-137181748 on Build GRCh38
rs760638299 Chr.9: 137181874-137181875 on Build GRCh38
rs111289442 Chr.9: 137181870-137181870 on Build GRCh38
rs868642760 Chr.9: 137181965-137181965 on Build GRCh38
rs139552269 Chr.9: 137181804-137181804 on Build GRCh38
rs942108056 Chr.9: 137182003-137182003 on Build GRCh38
rs757162417 Chr.9: 137181634-137181634 on Build GRCh38
rs1056618738 Chr.9: 137181766-137181766 on Build GRCh38
rs776840253 Chr.9: 137181924-137181924 on Build GRCh38
rs753903946 Chr.9: 137181638-137181638 on Build GRCh38
rs987970811 Chr.9: 137181873-137181873 on Build GRCh38
rs781632508 Chr.9: 137181847-137181847 on Build GRCh38
rs930399951 Chr.9: 137181960-137181960 on Build GRCh38
rs946732888 Chr.9: 137181687-137181687 on Build GRCh38
rs760428991 Chr.9: 137181886-137181886 on Build GRCh38
rs373745299 Chr.9: 137181612-137181612 on Build GRCh38
rs898947894 Chr.9: 137181793-137181793 on Build GRCh38
rs1048236157 Chr.9: 137181761-137181761 on Build GRCh38
rs866845913 Chr.9: 137181689-137181689 on Build GRCh38
rs1055563238 Chr.9: 137182007-137182007 on Build GRCh38
rs28651923 Chr.9: 137181909-137181909 on Build GRCh38
rs888783671 Chr.9: 137181976-137181976 on Build GRCh38
rs778023486 Chr.9: 137181652-137181652 on Build GRCh38
rs569174858 Chr.9: 137181625-137181625 on Build GRCh38
rs769428566 Chr.9: 137181665-137181665 on Build GRCh38
rs929394579 Chr.9: 137181736-137181736 on Build GRCh38
rs749783072 Chr.9: 137181992-137181992 on Build GRCh38
rs778022128 Chr.9: 137181755-137181755 on Build GRCh38
rs542809566 Chr.9: 137181907-137181907 on Build GRCh38
rs72763283 Chr.9: 137182012-137182012 on Build GRCh38
rs768234499 Chr.9: 137181790-137181790 on Build GRCh38
rs567416919 Chr.9: 137181973-137181973 on Build GRCh38
rs373925611 Chr.9: 137181825-137181826 on Build GRCh38
rs1050069744 Chr.9: 137181971-137181971 on Build GRCh38
rs909327556 Chr.9: 137181994-137181994 on Build GRCh38
rs761572272 Chr.9: 137181802-137181802 on Build GRCh38
rs557011313 Chr.9: 137181621-137181621 on Build GRCh38
rs750298505 Chr.9: 137181642-137181643 on Build GRCh38
rs775165850 Chr.9: 137181772-137181772 on Build GRCh38
rs375903586 Chr.9: 137181703-137181703 on Build GRCh38
rs763793663 Chr.9: 137181684-137181684 on Build GRCh38
rs757624432 Chr.9: 137181760-137181760 on Build GRCh38
rs781656030 Chr.9: 137181763-137181763 on Build GRCh38
rs780765550 Chr.9: 137181911-137181911 on Build GRCh38
rs751797577 Chr.9: 137181830-137181830 on Build GRCh38
rs370895377 Chr.9: 137181759-137181759 on Build GRCh38
rs187170492 Chr.9: 137181940-137181940 on Build GRCh38
rs975974338 Chr.9: 137181937-137181937 on Build GRCh38
rs774033779 Chr.9: 137181798-137181798 on Build GRCh38
rs923035219 Chr.9: 137181815-137181815 on Build GRCh38
rs373230905 Chr.9: 137181664-137181664 on Build GRCh38
rs757359561 Chr.9: 137181841-137181841 on Build GRCh38
rs746188952 Chr.9: 137181853-137181853 on Build GRCh38
rs566917627 Chr.9: 137182016-137182016 on Build GRCh38
rs73571572 Chr.9: 137181654-137181654 on Build GRCh38
rs758648523 Chr.9: 137181752-137181752 on Build GRCh38
rs957219907 Chr.9: 137181645-137181645 on Build GRCh38
rs371722410 Chr.9: 137181905-137181905 on Build GRCh38
rs551357840 Chr.9: 137181912-137181912 on Build GRCh38
rs28665875 Chr.9: 137181987-137181987 on Build GRCh38
rs772824308 Chr.9: 137181807-137181807 on Build GRCh38
rs773020889 Chr.9: 137181885-137181885 on Build GRCh38
rs376312292 Chr.9: 137181640-137181640 on Build GRCh38
rs775535887 Chr.9: 137181988-137181988 on Build GRCh38
rs764277676 Chr.9: 137181825-137181825 on Build GRCh38
rs780677757 Chr.9: 137181861-137181861 on Build GRCh38
rs762711070 Chr.9: 137181681-137181681 on Build GRCh38
rs777177726 Chr.9: 137181659-137181659 on Build GRCh38
rs887751750 Chr.9: 137181601-137181601 on Build GRCh38
rs376778830 Chr.9: 137181620-137181620 on Build GRCh38
rs539341059 Chr.9: 137182043-137182043 on Build GRCh38
rs1034313815 Chr.9: 137181615-137181615 on Build GRCh38
rs953439578 Chr.9: 137181610-137181610 on Build GRCh38
rs755978183 Chr.9: 137181723-137181723 on Build GRCh38
rs1008788924 Chr.9: 137181866-137181866 on Build GRCh38
rs768199316 Chr.9: 137181867-137181867 on Build GRCh38
rs150053006 Chr.9: 137181820-137181820 on Build GRCh38
rs748858129 Chr.9: 137181865-137181865 on Build GRCh38
rs771384409 Chr.9: 137181658-137181658 on Build GRCh38
rs956765336 Chr.9: 137181624-137181624 on Build GRCh38
rs553438864 Chr.9: 137181688-137181688 on Build GRCh38
rs142080330 Chr.9: 137181744-137181744 on Build GRCh38
rs966824466 Chr.9: 137181893-137181893 on Build GRCh38
rs779335194 Chr.9: 137182041-137182041 on Build GRCh38
rs747850425 Chr.9: 137181874-137181874 on Build GRCh38
rs28369482 Chr.9: 137181906-137181906 on Build GRCh38
rs146295520 Chr.9: 137181779-137181779 on Build GRCh38
rs923072147 Chr.9: 137181670-137181670 on Build GRCh38
rs766263225 Chr.9: 137181812-137181812 on Build GRCh38
rs563358017 Chr.9: 137181898-137181898 on Build GRCh38
rs929089904 Chr.9: 137181975-137181975 on Build GRCh38
rs145419936 Chr.9: 137181859-137181859 on Build GRCh38
rs774046730 Chr.9: 137181697-137181697 on Build GRCh38
rs987251006 Chr.9: 137181611-137181611 on Build GRCh38
rs538103729 Chr.9: 137181706-137181706 on Build GRCh38
rs924317463 Chr.9: 137181643-137181643 on Build GRCh38
rs376190893 Chr.9: 137181884-137181884 on Build GRCh38
rs892424906 Chr.9: 137182011-137182011 on Build GRCh38
rs376155568 Chr.9: 137181641-137181641 on Build GRCh38
rs376993478 Chr.9: 137181808-137181808 on Build GRCh38
rs775077992 Chr.9: 137181668-137181668 on Build GRCh38
rs751308403 Chr.9: 137181630-137181630 on Build GRCh38
rs550699184 Chr.9: 137181961-137181961 on Build GRCh38
rs373578381 Chr.9: 137181771-137181771 on Build GRCh38
rs765313489 Chr.9: 137181732-137181732 on Build GRCh38
rs368025140 Chr.9: 137181908-137181908 on Build GRCh38
rs781142743 Chr.9: 137181637-137181637 on Build GRCh38
rs746385223 Chr.9: 137181769-137181769 on Build GRCh38
rs58384967 Chr.9: 137181894-137181894 on Build GRCh38
rs150695160 Chr.9: 137181910-137181910 on Build GRCh38

View Full Product Details