Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137347879-137348376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD3
Entrez Gene ID
54932
Gene Name
exonuclease 3'-5' domain containing 3
Gene Aliases
mut-7
UniGene
Hs.495553
Gene Chromosome Location
Chr.9: 137306894-137423262 on Build GRCh38
Transcripts
NM_017820.4 XM_011518810.1 XM_011518814.1 XM_011518815.1 XM_017014861.1 XM_017014862.1 XM_017014863.1 XM_017014864.1 XM_017014865.1 XM_017014866.1 XM_017014867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3089216 c.1998+10C>T p.? Chr.9: 137348061-137348061 on Build GRCh38
6039569 c.1495-251_1831-40del1505 p.? Chr.9: 137348278-137349782 on Build GRCh38
389725 c.1892G>T p.R631L Chr.9: 137348177-137348177 on Build GRCh38
3848116 c.1927C>T p.R643W Chr.9: 137348142-137348142 on Build GRCh38
5137556 c.1984C>T p.R662C Chr.9: 137348085-137348085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117681406 Chr.9: 137348251-137348251 on Build GRCh38
rs543431814 Chr.9: 137347912-137347912 on Build GRCh38
rs201090340 Chr.9: 137348240-137348240 on Build GRCh38
rs1051745949 Chr.9: 137348052-137348052 on Build GRCh38
rs767343485 Chr.9: 137348151-137348151 on Build GRCh38
rs533338047 Chr.9: 137347903-137347903 on Build GRCh38
rs780140324 Chr.9: 137348118-137348118 on Build GRCh38
rs745772775 Chr.9: 137348032-137348032 on Build GRCh38
rs117446810 Chr.9: 137348321-137348321 on Build GRCh38
rs200498331 Chr.9: 137348084-137348084 on Build GRCh38
rs377169281 Chr.9: 137348331-137348331 on Build GRCh38
rs529291876 Chr.9: 137348306-137348306 on Build GRCh38
rs758878437 Chr.9: 137348276-137348277 on Build GRCh38
rs765802505 Chr.9: 137348078-137348078 on Build GRCh38
rs202080579 Chr.9: 137348077-137348077 on Build GRCh38
rs941865177 Chr.9: 137347933-137347933 on Build GRCh38
rs749269507 Chr.9: 137348170-137348170 on Build GRCh38
rs893974857 Chr.9: 137348082-137348082 on Build GRCh38
rs780097636 Chr.9: 137348276-137348276 on Build GRCh38
rs377069829 Chr.9: 137348188-137348188 on Build GRCh38
rs1004529707 Chr.9: 137348341-137348341 on Build GRCh38
rs547056556 Chr.9: 137348061-137348061 on Build GRCh38
rs765675666 Chr.9: 137348138-137348138 on Build GRCh38
rs368707251 Chr.9: 137347930-137347930 on Build GRCh38
rs749448873 Chr.9: 137348277-137348277 on Build GRCh38
rs367737146 Chr.9: 137348124-137348124 on Build GRCh38
rs535339908 Chr.9: 137348149-137348149 on Build GRCh38
rs58941960 Chr.9: 137348274-137348274 on Build GRCh38
rs762239994 Chr.9: 137348209-137348209 on Build GRCh38
rs376408986 Chr.9: 137348062-137348062 on Build GRCh38
rs563738859 Chr.9: 137347930-137347930 on Build GRCh38
rs756977327 Chr.9: 137348109-137348109 on Build GRCh38
rs776689407 Chr.9: 137348132-137348132 on Build GRCh38
rs769898285 Chr.9: 137348042-137348042 on Build GRCh38
rs372777188 Chr.9: 137348193-137348193 on Build GRCh38
rs767216999 Chr.9: 137348216-137348216 on Build GRCh38
rs760759416 Chr.9: 137348063-137348063 on Build GRCh38
rs752379047 Chr.9: 137348255-137348255 on Build GRCh38
rs367634582 Chr.9: 137348060-137348060 on Build GRCh38
rs200395995 Chr.9: 137348140-137348140 on Build GRCh38
rs35170062 Chr.9: 137348027-137348027 on Build GRCh38
rs4078069 Chr.9: 137348182-137348182 on Build GRCh38
rs931313196 Chr.9: 137348301-137348301 on Build GRCh38
rs61677362 Chr.9: 137348320-137348320 on Build GRCh38
rs200930714 Chr.9: 137348125-137348125 on Build GRCh38
rs28671254 Chr.9: 137348112-137348112 on Build GRCh38
rs1002428367 Chr.9: 137348178-137348178 on Build GRCh38
rs770993858 Chr.9: 137348184-137348184 on Build GRCh38
rs754981932 Chr.9: 137348177-137348177 on Build GRCh38
rs781353221 Chr.9: 137348269-137348269 on Build GRCh38
rs148957448 Chr.9: 137348116-137348116 on Build GRCh38
rs980609664 Chr.9: 137347998-137347998 on Build GRCh38
rs376214483 Chr.9: 137348141-137348141 on Build GRCh38
rs529554307 Chr.9: 137347936-137347936 on Build GRCh38
rs780089309 Chr.9: 137348164-137348164 on Build GRCh38
rs969203646 Chr.9: 137347977-137347977 on Build GRCh38
rs775409846 Chr.9: 137348055-137348055 on Build GRCh38
rs749030027 Chr.9: 137348122-137348122 on Build GRCh38
rs987342933 Chr.9: 137348267-137348267 on Build GRCh38
rs770374323 Chr.9: 137348026-137348026 on Build GRCh38
rs756843662 Chr.9: 137348150-137348150 on Build GRCh38
rs759619304 Chr.9: 137348133-137348133 on Build GRCh38
rs763757887 Chr.9: 137348085-137348085 on Build GRCh38
rs967697682 Chr.9: 137348213-137348214 on Build GRCh38
rs200202616 Chr.9: 137348208-137348208 on Build GRCh38
rs760221601 Chr.9: 137348229-137348229 on Build GRCh38
rs762903734 Chr.9: 137348058-137348058 on Build GRCh38
rs145788389 Chr.9: 137348029-137348029 on Build GRCh38
rs768974173 Chr.9: 137348282-137348282 on Build GRCh38
rs780547676 Chr.9: 137348111-137348111 on Build GRCh38
rs746062681 Chr.9: 137348272-137348272 on Build GRCh38
rs202048990 Chr.9: 137348073-137348073 on Build GRCh38
rs185180968 Chr.9: 137347973-137347973 on Build GRCh38
rs774933960 Chr.9: 137348287-137348287 on Build GRCh38
rs563531041 Chr.9: 137348304-137348304 on Build GRCh38
rs368003193 Chr.9: 137348155-137348155 on Build GRCh38
rs765955903 Chr.9: 137348235-137348235 on Build GRCh38
rs1044401898 Chr.9: 137348303-137348303 on Build GRCh38
rs771233229 Chr.9: 137348126-137348126 on Build GRCh38
rs772341387 Chr.9: 137348223-137348223 on Build GRCh38
rs769266306 Chr.9: 137348207-137348207 on Build GRCh38
rs753436084 Chr.9: 137348238-137348238 on Build GRCh38
rs377232401 Chr.9: 137348268-137348268 on Build GRCh38
rs983346664 Chr.9: 137347910-137347910 on Build GRCh38
rs751206766 Chr.9: 137348102-137348102 on Build GRCh38
rs118030525 Chr.9: 137348192-137348192 on Build GRCh38
rs764339032 Chr.9: 137348142-137348142 on Build GRCh38
rs748043355 Chr.9: 137348183-137348183 on Build GRCh38
rs558014887 Chr.9: 137348220-137348220 on Build GRCh38
rs867434191 Chr.9: 137348107-137348107 on Build GRCh38
rs563506690 Chr.9: 137347924-137347924 on Build GRCh38
rs1051795837 Chr.9: 137348233-137348233 on Build GRCh38
rs143397897 Chr.9: 137348335-137348336 on Build GRCh38
rs776730395 Chr.9: 137348074-137348074 on Build GRCh38
rs927870574 Chr.9: 137348002-137348002 on Build GRCh38
rs781146880 Chr.9: 137348031-137348031 on Build GRCh38
rs755695023 Chr.9: 137348160-137348160 on Build GRCh38
rs757938194 Chr.9: 137347911-137347911 on Build GRCh38
rs147058366 Chr.9: 137348217-137348218 on Build GRCh38
rs995565938 Chr.9: 137347927-137347927 on Build GRCh38
rs774800630 Chr.9: 137348294-137348294 on Build GRCh38
rs199557029 Chr.9: 137348191-137348191 on Build GRCh38

View Full Product Details