Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137351148-137351674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD3
Entrez Gene ID
54932
Gene Name
exonuclease 3'-5' domain containing 3
Gene Aliases
mut-7
UniGene
Hs.495553
Gene Chromosome Location
Chr.9: 137306894-137423262 on Build GRCh38
Transcripts
NM_017820.4 XM_011518810.1 XM_011518814.1 XM_011518815.1 XM_017014861.1 XM_017014862.1 XM_017014863.1 XM_017014864.1 XM_017014865.1 XM_017014866.1 XM_017014867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5176521 c.1174-7C>T p.? Chr.9: 137351535-137351535 on Build GRCh38
5733514 c.1273G>A p.V425M Chr.9: 137351429-137351429 on Build GRCh38
1461388 c.1238G>A p.R413Q Chr.9: 137351464-137351464 on Build GRCh38
6034982 c.1255G>T p.V419L Chr.9: 137351447-137351447 on Build GRCh38
7222714 c.1362G>A p.S454S Chr.9: 137351340-137351340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548116967 Chr.9: 137351470-137351470 on Build GRCh38
rs765883082 Chr.9: 137351418-137351418 on Build GRCh38
rs952595103 Chr.9: 137351265-137351265 on Build GRCh38
rs750623889 Chr.9: 137351450-137351450 on Build GRCh38
rs374214809 Chr.9: 137351471-137351471 on Build GRCh38
rs548894905 Chr.9: 137351465-137351465 on Build GRCh38
rs1017701585 Chr.9: 137351489-137351489 on Build GRCh38
rs1016454499 Chr.9: 137351626-137351626 on Build GRCh38
rs376504398 Chr.9: 137351533-137351533 on Build GRCh38
rs757405748 Chr.9: 137351375-137351375 on Build GRCh38
rs776089638 Chr.9: 137351291-137351291 on Build GRCh38
rs779247850 Chr.9: 137351400-137351400 on Build GRCh38
rs376002483 Chr.9: 137351194-137351194 on Build GRCh38
rs761601756 Chr.9: 137351168-137351168 on Build GRCh38
rs764088188 Chr.9: 137351429-137351434 on Build GRCh38
rs11534416 Chr.9: 137351598-137351598 on Build GRCh38
rs754912457 Chr.9: 137351362-137351362 on Build GRCh38
rs377673439 Chr.9: 137351244-137351244 on Build GRCh38
rs537984909 Chr.9: 137351581-137351581 on Build GRCh38
rs770992060 Chr.9: 137351627-137351627 on Build GRCh38
rs377284984 Chr.9: 137351348-137351348 on Build GRCh38
rs1012737464 Chr.9: 137351208-137351208 on Build GRCh38
rs967422040 Chr.9: 137351553-137351553 on Build GRCh38
rs201793473 Chr.9: 137351464-137351464 on Build GRCh38
rs756335375 Chr.9: 137351461-137351461 on Build GRCh38
rs983891044 Chr.9: 137351269-137351269 on Build GRCh38
rs750056988 Chr.9: 137351364-137351364 on Build GRCh38
rs574380264 Chr.9: 137351222-137351222 on Build GRCh38
rs769667460 Chr.9: 137351508-137351508 on Build GRCh38
rs113473403 Chr.9: 137351414-137351414 on Build GRCh38
rs1018051823 Chr.9: 137351295-137351295 on Build GRCh38
rs773753469 Chr.9: 137351473-137351473 on Build GRCh38
rs767110521 Chr.9: 137351173-137351173 on Build GRCh38
rs994107170 Chr.9: 137351243-137351243 on Build GRCh38
rs895262595 Chr.9: 137351199-137351199 on Build GRCh38
rs374971103 Chr.9: 137351351-137351351 on Build GRCh38
rs976811075 Chr.9: 137351535-137351535 on Build GRCh38
rs767684381 Chr.9: 137351444-137351444 on Build GRCh38
rs200551018 Chr.9: 137351442-137351442 on Build GRCh38
rs775434946 Chr.9: 137351513-137351513 on Build GRCh38
rs559869849 Chr.9: 137351313-137351313 on Build GRCh38
rs760064563 Chr.9: 137351415-137351415 on Build GRCh38
rs369649398 Chr.9: 137351290-137351290 on Build GRCh38
rs562865667 Chr.9: 137351357-137351357 on Build GRCh38
rs780034262 Chr.9: 137351393-137351393 on Build GRCh38
rs762914549 Chr.9: 137351519-137351519 on Build GRCh38
rs781408432 Chr.9: 137351379-137351379 on Build GRCh38
rs1025943468 Chr.9: 137351245-137351245 on Build GRCh38
rs1011990318 Chr.9: 137351198-137351198 on Build GRCh38
rs1046146159 Chr.9: 137351422-137351422 on Build GRCh38
rs964844705 Chr.9: 137351330-137351330 on Build GRCh38
rs201064460 Chr.9: 137351441-137351441 on Build GRCh38
rs750007244 Chr.9: 137351175-137351175 on Build GRCh38
rs759512243 Chr.9: 137351430-137351430 on Build GRCh38
rs997678239 Chr.9: 137351550-137351550 on Build GRCh38
rs753517574 Chr.9: 137351185-137351185 on Build GRCh38
rs926172805 Chr.9: 137351374-137351374 on Build GRCh38
rs374728920 Chr.9: 137351178-137351178 on Build GRCh38
rs115216154 Chr.9: 137351192-137351192 on Build GRCh38
rs749776822 Chr.9: 137351394-137351394 on Build GRCh38
rs777818090 Chr.9: 137351193-137351193 on Build GRCh38
rs774327881 Chr.9: 137351444-137351445 on Build GRCh38
rs766336829 Chr.9: 137351195-137351195 on Build GRCh38
rs963997395 Chr.9: 137351642-137351642 on Build GRCh38
rs369865833 Chr.9: 137351373-137351373 on Build GRCh38
rs762856317 Chr.9: 137351366-137351366 on Build GRCh38
rs923452680 Chr.9: 137351345-137351345 on Build GRCh38
rs117448792 Chr.9: 137351634-137351634 on Build GRCh38
rs774900550 Chr.9: 137351337-137351337 on Build GRCh38
rs755379073 Chr.9: 137351396-137351396 on Build GRCh38
rs144883989 Chr.9: 137351522-137351522 on Build GRCh38
rs879036579 Chr.9: 137351459-137351459 on Build GRCh38
rs755133091 Chr.9: 137351566-137351566 on Build GRCh38
rs745952444 Chr.9: 137351289-137351289 on Build GRCh38
rs371400451 Chr.9: 137351176-137351176 on Build GRCh38
rs1031304786 Chr.9: 137351278-137351278 on Build GRCh38
rs761540359 Chr.9: 137351339-137351339 on Build GRCh38
rs761677018 Chr.9: 137351541-137351541 on Build GRCh38
rs1030118788 Chr.9: 137351543-137351543 on Build GRCh38
rs373734360 Chr.9: 137351640-137351640 on Build GRCh38
rs916115155 Chr.9: 137351621-137351621 on Build GRCh38
rs746651357 Chr.9: 137351492-137351492 on Build GRCh38
rs751364351 Chr.9: 137351531-137351531 on Build GRCh38
rs776138692 Chr.9: 137351503-137351503 on Build GRCh38
rs756246616 Chr.9: 137351382-137351382 on Build GRCh38
rs781245978 Chr.9: 137351268-137351268 on Build GRCh38
rs779579123 Chr.9: 137351469-137351469 on Build GRCh38
rs373425377 Chr.9: 137351380-137351380 on Build GRCh38
rs200494966 Chr.9: 137351504-137351504 on Build GRCh38
rs922663676 Chr.9: 137351334-137351334 on Build GRCh38
rs571020522 Chr.9: 137351474-137351474 on Build GRCh38
rs201801336 Chr.9: 137351401-137351401 on Build GRCh38
rs991737557 Chr.9: 137351599-137351599 on Build GRCh38
rs770556244 Chr.9: 137351498-137351498 on Build GRCh38
rs369943946 Chr.9: 137351227-137351228 on Build GRCh38
rs753844140 Chr.9: 137351361-137351361 on Build GRCh38
rs1024668976 Chr.9: 137351220-137351220 on Build GRCh38
rs58444859 Chr.9: 137351340-137351340 on Build GRCh38
rs545668606 Chr.9: 137351234-137351234 on Build GRCh38
rs765162423 Chr.9: 137351432-137351432 on Build GRCh38
rs528788626 Chr.9: 137351218-137351218 on Build GRCh38
rs977794765 Chr.9: 137351555-137351555 on Build GRCh38
rs754852116 Chr.9: 137351300-137351300 on Build GRCh38
rs778273275 Chr.9: 137351532-137351532 on Build GRCh38
rs771201636 Chr.9: 137351197-137351197 on Build GRCh38
rs760243505 Chr.9: 137351341-137351341 on Build GRCh38
rs1042352134 Chr.9: 137351172-137351172 on Build GRCh38
rs765004634 Chr.9: 137351363-137351363 on Build GRCh38
rs976804642 Chr.9: 137351326-137351326 on Build GRCh38
rs374379013 Chr.9: 137351235-137351235 on Build GRCh38
rs372891499 Chr.9: 137351429-137351429 on Build GRCh38
rs371027809 Chr.9: 137351558-137351558 on Build GRCh38
rs532185052 Chr.9: 137351325-137351325 on Build GRCh38

View Full Product Details