Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECAB1
Entrez Gene ID
64168
Gene Name
N-terminal EF-hand calcium binding protein 1
Gene Aliases
EFCBP1, STIP-1
Chromosome Location
Chr.8: 90791693 - 90959402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022351.4 NP_071746.1 11 - 12 1135 146
GenBank mRNA AF193756.1 - 7 - 8 653 146
AF414126.2 - 12 - 13 1224 146
AK095533.1 - 11 - 12 1048 146
AY224210.1 - 11 - 12 941 146
BC016340.1 - 2 - 3 245 146
BC068448.1 - 11 - 12 1048 146

View Full Product Details