The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033105.4 | NP_149096.2 | 1 - 2 | 146 | 71 |
XM_011517620.2 | XP_011515922.1 | 1 - 2 | 139 | 71 | |
GenBank mRNA | AF368276.1 | - | 1 - 2 | 124 | 71 |
AL133660.1 | - | 1 - 2 | 88 | 71 | |
BC016742.2 | - | 1 - 2 | 143 | 71 | |
BG701336.1 | - | 1 - 2 | 161 | 71 |