Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
Chromosome Location
Chr.4: 119296419 - 119304445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001701.3 NP_001001701.2 1 - 2 474 93
NM_001170330.1 NP_001163801.1 2 - 3 595 93
GenBank mRNA AF075061.1 - 1 - 2 282 93
AK025703.1 - 1 - 2 238 93
BC017399.1 - 1 - 2 297 93
DA353224.1 - 1 - 2 474 93

View Full Product Details