The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000302.3 | NP_000293.2 | 8 - 9 | 954 | 78 |
NM_001316320.1 | NP_001303249.1 | 9 - 10 | 1095 | 78 | |
GenBank mRNA | AK294688.1 | - | 8 - 9 | 785 | 78 |
AK299150.1 | - | 9 - 10 | 1028 | 78 | |
AK312243.1 | - | 8 - 9 | 887 | 78 | |
AK316285.1 | - | 9 - 10 | 1031 | 78 | |
BC016657.2 | - | 8 - 9 | 858 | 78 | |
M98252.1 | - | 8 - 9 | 865 | 78 |