The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000696.3 | NP_000687.3 | 9 - 10 | 1449 | 101 |
XM_011509294.2 | XP_011507596.1 | 9 - 10 | 1513 | 101 | |
GenBank mRNA | AF172093.1 | - | 9 - 10 | 1272 | 101 |
AK293520.1 | - | 9 - 10 | 1353 | 101 | |
AK302183.1 | - | 9 - 10 | 1280 | 101 | |
AK302191.1 | - | 9 - 10 | 1222 | 101 | |
AK312751.1 | - | 9 - 10 | 1378 | 101 | |
BC070030.1 | - | 9 - 10 | 1378 | 101 | |
BC151140.1 | - | 9 - 10 | 1437 | 101 | |
BC151141.1 | - | 9 - 10 | 1437 | 101 | |
U34252.1 | - | 9 - 10 | 1646 | 101 | |
U50203.1 | - | 9 - 10 | 1188 | 101 | |
X75425.1 | - | 8 - 9 | 1176 | 101 |