Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50193013-50193509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP4
Entrez Gene ID
8852
Gene Name
A-kinase anchoring protein 4
Gene Aliases
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
UniGene
Hs.97633
Gene Chromosome Location
Chr.X: 50190768-50201013 on Build GRCh38
Transcripts
NM_003886.2 NM_139289.1

SNP Detail

SNP ID Chromosome Location
rs187265787 Chr.X: 50193166-50193166 on Build GRCh38
rs143420996 Chr.X: 50193204-50193204 on Build GRCh38
rs1132159 Chr.X: 50193128-50193128 on Build GRCh38
rs201649248 Chr.X: 50193038-50193038 on Build GRCh38
rs139049075 Chr.X: 50193055-50193055 on Build GRCh38
rs781999489 Chr.X: 50193224-50193224 on Build GRCh38
rs782817329 Chr.X: 50193372-50193372 on Build GRCh38
rs782620501 Chr.X: 50193413-50193413 on Build GRCh38
rs144194103 Chr.X: 50193445-50193445 on Build GRCh38
rs782184143 Chr.X: 50193258-50193258 on Build GRCh38
rs1055930555 Chr.X: 50193217-50193217 on Build GRCh38
rs143933731 Chr.X: 50193064-50193064 on Build GRCh38
rs781884424 Chr.X: 50193133-50193133 on Build GRCh38
rs267606473 Chr.X: 50193105-50193105 on Build GRCh38
rs930232562 Chr.X: 50193071-50193071 on Build GRCh38
rs782357100 Chr.X: 50193282-50193282 on Build GRCh38
rs139438809 Chr.X: 50193318-50193318 on Build GRCh38
rs781814079 Chr.X: 50193396-50193396 on Build GRCh38
rs782131534 Chr.X: 50193297-50193297 on Build GRCh38
rs1132160 Chr.X: 50193119-50193119 on Build GRCh38
rs782214760 Chr.X: 50193420-50193420 on Build GRCh38
rs782641171 Chr.X: 50193240-50193240 on Build GRCh38
rs782502351 Chr.X: 50193153-50193153 on Build GRCh38
rs781869716 Chr.X: 50193140-50193140 on Build GRCh38
rs367661119 Chr.X: 50193294-50193294 on Build GRCh38
rs148654052 Chr.X: 50193117-50193117 on Build GRCh38
rs782437666 Chr.X: 50193407-50193407 on Build GRCh38
rs782464382 Chr.X: 50193441-50193441 on Build GRCh38
rs782149049 Chr.X: 50193346-50193346 on Build GRCh38
rs782458932 Chr.X: 50193239-50193239 on Build GRCh38
rs146755588 Chr.X: 50193241-50193241 on Build GRCh38
rs202187814 Chr.X: 50193315-50193315 on Build GRCh38
rs143954829 Chr.X: 50193079-50193079 on Build GRCh38
rs200249971 Chr.X: 50193427-50193427 on Build GRCh38
rs782746899 Chr.X: 50193129-50193129 on Build GRCh38
rs781961150 Chr.X: 50193043-50193043 on Build GRCh38
rs782146975 Chr.X: 50193225-50193225 on Build GRCh38
rs781919108 Chr.X: 50193104-50193104 on Build GRCh38
rs998778865 Chr.X: 50193367-50193367 on Build GRCh38
rs782713046 Chr.X: 50193366-50193366 on Build GRCh38
rs375412934 Chr.X: 50193391-50193391 on Build GRCh38
rs928823753 Chr.X: 50193161-50193161 on Build GRCh38
rs781940118 Chr.X: 50193255-50193255 on Build GRCh38
rs782288244 Chr.X: 50193444-50193444 on Build GRCh38
rs782044480 Chr.X: 50193078-50193078 on Build GRCh38
rs782812592 Chr.X: 50193187-50193187 on Build GRCh38
rs781869477 Chr.X: 50193249-50193249 on Build GRCh38
rs372025415 Chr.X: 50193428-50193428 on Build GRCh38
rs781886271 Chr.X: 50193202-50193202 on Build GRCh38
rs782552330 Chr.X: 50193218-50193218 on Build GRCh38
rs782091014 Chr.X: 50193380-50193380 on Build GRCh38
rs782675260 Chr.X: 50193392-50193392 on Build GRCh38
rs190004634 Chr.X: 50193101-50193101 on Build GRCh38
rs201779837 Chr.X: 50193446-50193446 on Build GRCh38

View Full Product Details