Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50196785-50197304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP4
Entrez Gene ID
8852
Gene Name
A-kinase anchoring protein 4
Gene Aliases
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
UniGene
Hs.97633
Gene Chromosome Location
Chr.X: 50190768-50201013 on Build GRCh38
Transcripts
NM_003886.2 NM_139289.1

SNP Detail

SNP ID Chromosome Location
rs782579748 Chr.X: 50197256-50197256 on Build GRCh38
rs960628837 Chr.X: 50197077-50197077 on Build GRCh38
rs146253420 Chr.X: 50196932-50196932 on Build GRCh38
rs782207677 Chr.X: 50197078-50197078 on Build GRCh38
rs112285697 Chr.X: 50196996-50196996 on Build GRCh38
rs139319738 Chr.X: 50196977-50196977 on Build GRCh38
rs782037617 Chr.X: 50196992-50196992 on Build GRCh38
rs979295415 Chr.X: 50197233-50197233 on Build GRCh38
rs17174079 Chr.X: 50196819-50196819 on Build GRCh38
rs782661882 Chr.X: 50196811-50196811 on Build GRCh38
rs782502960 Chr.X: 50196967-50196967 on Build GRCh38
rs143128429 Chr.X: 50197159-50197159 on Build GRCh38
rs376665487 Chr.X: 50196843-50196843 on Build GRCh38
rs186535169 Chr.X: 50197261-50197261 on Build GRCh38
rs375169998 Chr.X: 50197154-50197154 on Build GRCh38
rs368770403 Chr.X: 50196883-50196883 on Build GRCh38
rs782293667 Chr.X: 50196884-50196884 on Build GRCh38
rs782376800 Chr.X: 50196989-50196989 on Build GRCh38
rs969614882 Chr.X: 50197160-50197160 on Build GRCh38
rs781915113 Chr.X: 50197013-50197013 on Build GRCh38
rs369965111 Chr.X: 50197024-50197024 on Build GRCh38
rs190421799 Chr.X: 50196940-50196940 on Build GRCh38
rs900685100 Chr.X: 50196829-50196829 on Build GRCh38
rs782687013 Chr.X: 50196939-50196939 on Build GRCh38
rs374649887 Chr.X: 50196995-50196995 on Build GRCh38
rs200799849 Chr.X: 50196904-50196904 on Build GRCh38
rs782063466 Chr.X: 50196856-50196856 on Build GRCh38
rs782027194 Chr.X: 50197238-50197238 on Build GRCh38
rs782273178 Chr.X: 50196988-50196988 on Build GRCh38
rs182053672 Chr.X: 50197063-50197063 on Build GRCh38
rs782456419 Chr.X: 50196926-50196926 on Build GRCh38
rs782749208 Chr.X: 50196831-50196831 on Build GRCh38
rs781841685 Chr.X: 50196906-50196906 on Build GRCh38
rs782669322 Chr.X: 50196948-50196948 on Build GRCh38
rs926027382 Chr.X: 50197251-50197251 on Build GRCh38
rs782307694 Chr.X: 50197250-50197250 on Build GRCh38
rs782729100 Chr.X: 50197025-50197025 on Build GRCh38
rs782180406 Chr.X: 50196957-50196957 on Build GRCh38

View Full Product Details