The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001749.3 | NP_001740.1 | 1 - 2 | 230 | 77 |
GenBank mRNA | AK289606.1 | - | 1 - 2 | 169 | 77 |
BC000592.2 | - | 1 - 2 | 48 | 77 | |
BC007779.2 | - | 1 - 2 | 58 | 77 | |
BC011903.2 | - | 1 - 2 | 48 | 77 | |
BQ898664.1 | - | 1 - 2 | 140 | 77 | |
DR759763.1 | - | 1 - 2 | 230 | 77 | |
X04106.1 | - | 1 - 2 | 141 | 77 |