Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55006277-55006777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APEX2
Entrez Gene ID
27301
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
Gene Aliases
APE2, APEXL2, XTH2, ZGRF2
UniGene
Hs.659558
Gene Chromosome Location
Chr.X: 55000323-55007873 on Build GRCh38
Transcripts
NM_001271748.1 NM_014481.3

SNP Detail

SNP ID Chromosome Location
rs960257841 Chr.X: 55006314-55006314 on Build GRCh38
rs1023556581 Chr.X: 55006323-55006323 on Build GRCh38
rs142257625 Chr.X: 55006713-55006713 on Build GRCh38
rs931544803 Chr.X: 55006644-55006644 on Build GRCh38
rs868705455 Chr.X: 55006681-55006681 on Build GRCh38
rs187479006 Chr.X: 55006542-55006542 on Build GRCh38
rs765983392 Chr.X: 55006677-55006677 on Build GRCh38
rs751057658 Chr.X: 55006484-55006484 on Build GRCh38
rs1014801737 Chr.X: 55006723-55006723 on Build GRCh38
rs754472880 Chr.X: 55006497-55006497 on Build GRCh38
rs758037914 Chr.X: 55006468-55006468 on Build GRCh38
rs145295903 Chr.X: 55006595-55006595 on Build GRCh38
rs145122391 Chr.X: 55006683-55006683 on Build GRCh38
rs758761322 Chr.X: 55006481-55006482 on Build GRCh38
rs778186956 Chr.X: 55006506-55006506 on Build GRCh38
rs750220870 Chr.X: 55006668-55006668 on Build GRCh38
rs762764908 Chr.X: 55006676-55006676 on Build GRCh38
rs771481521 Chr.X: 55006547-55006547 on Build GRCh38
rs754465359 Chr.X: 55006712-55006712 on Build GRCh38
rs769497829 Chr.X: 55006373-55006373 on Build GRCh38
rs200406687 Chr.X: 55006559-55006559 on Build GRCh38
rs748749797 Chr.X: 55006397-55006397 on Build GRCh38
rs968368478 Chr.X: 55006341-55006341 on Build GRCh38
rs746169749 Chr.X: 55006567-55006567 on Build GRCh38
rs772853512 Chr.X: 55006374-55006374 on Build GRCh38
rs765080268 Chr.X: 55006614-55006614 on Build GRCh38
rs952906800 Chr.X: 55006435-55006435 on Build GRCh38
rs149134407 Chr.X: 55006691-55006691 on Build GRCh38
rs912285894 Chr.X: 55006591-55006591 on Build GRCh38
rs780742079 Chr.X: 55006692-55006692 on Build GRCh38
rs772439550 Chr.X: 55006730-55006730 on Build GRCh38
rs961043595 Chr.X: 55006577-55006577 on Build GRCh38
rs746256403 Chr.X: 55006729-55006729 on Build GRCh38
rs775731944 Chr.X: 55006594-55006594 on Build GRCh38
rs376410778 Chr.X: 55006406-55006406 on Build GRCh38
rs776595560 Chr.X: 55006604-55006604 on Build GRCh38
rs772195864 Chr.X: 55006583-55006583 on Build GRCh38
rs779505607 Chr.X: 55006725-55006725 on Build GRCh38
rs183348151 Chr.X: 55006470-55006470 on Build GRCh38
rs761722822 Chr.X: 55006610-55006610 on Build GRCh38
rs201475384 Chr.X: 55006501-55006501 on Build GRCh38
rs780178393 Chr.X: 55006743-55006743 on Build GRCh38

View Full Product Details