Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKS1
Entrez Gene ID
54903
Gene Name
Meckel syndrome, type 1
Gene Aliases
BBS13, MES, MKS, POC12
Chromosome Location
Chr.17: 58205436 - 58219605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165927.1 NP_001159399.1 4 - 5 482 81
NM_001321269.1 NP_001308198.1 4 - 5 493 81
NM_017777.3 NP_060247.2 4 - 5 493 81
XM_011524957.2 XP_011523259.1 4 - 5 473 81
XM_011524958.2 XP_011523260.1 4 - 5 473 81
XM_011524959.2 XP_011523261.1 4 - 5 472 81
XM_011524960.2 XP_011523262.1 4 - 5 472 81
XM_017024803.1 XP_016880292.1 4 - 5 441 81
XM_017024804.1 XP_016880293.1 4 - 5 441 81
GenBank mRNA AK000352.1 - 1 - 2 163 81
AK310815.1 - 4 - 5 482 81
BC010061.2 - 4 - 5 428 81
DQ185029.1 - 4 - 5 493 81

View Full Product Details