Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 148718949-148719487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF2
Entrez Gene ID
2334
Gene Name
AF4/FMR2 family member 2
Gene Aliases
FMR2, FMR2P, FRAXE, MRX2, OX19
UniGene
Hs.496911
Gene Chromosome Location
Chr.X: 148500619-149000663 on Build GRCh38
Transcripts
NM_001169122.1 NM_001169123.1 NM_001169124.1 NM_001169125.1 NM_001170628.1 NM_002025.3

SNP Detail

SNP ID Chromosome Location
rs62610426 Chr.X: 148719249-148719249 on Build GRCh38
rs782282910 Chr.X: 148719221-148719221 on Build GRCh38
rs184816699 Chr.X: 148719228-148719228 on Build GRCh38
rs140788054 Chr.X: 148719213-148719213 on Build GRCh38
rs184288076 Chr.X: 148718988-148718988 on Build GRCh38
rs782484350 Chr.X: 148719170-148719170 on Build GRCh38
rs1003203699 Chr.X: 148719301-148719301 on Build GRCh38
rs782443496 Chr.X: 148719225-148719225 on Build GRCh38
rs782373738 Chr.X: 148719097-148719097 on Build GRCh38
rs241084 Chr.X: 148719224-148719224 on Build GRCh38
rs147596059 Chr.X: 148719154-148719154 on Build GRCh38
rs986443520 Chr.X: 148719008-148719008 on Build GRCh38
rs782682054 Chr.X: 148719230-148719230 on Build GRCh38
rs188901204 Chr.X: 148719280-148719280 on Build GRCh38
rs782455987 Chr.X: 148719444-148719444 on Build GRCh38
rs942437989 Chr.X: 148719071-148719071 on Build GRCh38
rs782526454 Chr.X: 148719261-148719261 on Build GRCh38
rs782805821 Chr.X: 148719266-148719266 on Build GRCh38
rs781997605 Chr.X: 148719101-148719101 on Build GRCh38
rs189238334 Chr.X: 148719105-148719105 on Build GRCh38
rs1057106405 Chr.X: 148719453-148719453 on Build GRCh38
rs193231108 Chr.X: 148719155-148719155 on Build GRCh38
rs145217067 Chr.X: 148719011-148719011 on Build GRCh38

View Full Product Details