The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003849.3 | NP_003840.2 | 2 - 3 | 396 | 117 |
GenBank mRNA | AF104921.2 | - | 2 - 3 | 195 | 117 |
AK225737.1 | - | 2 - 3 | 173 | 117 | |
AK291082.1 | - | 2 - 3 | 195 | 117 | |
AK296741.1 | - | 2 - 3 | 203 | 117 | |
AK313290.1 | - | 2 - 3 | 175 | 117 | |
BC000504.2 | - | 2 - 3 | 199 | 117 | |
BM804835.1 | - | 2 - 3 | 382 | 117 | |
BP194715.1 | - | 2 - 3 | 397 | 117 | |
CR457139.1 | - | 2 - 3 | 164 | 117 | |
Z68204.1 | - | 2 - 3 | 170 | 117 |