Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location
Chr.13: 111114619 - 111305734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113511.2 NP_001106983.1 8 - 9 1826 102
NM_001113512.2 NP_001106984.1 6 - 7 1676 102
NM_001113513.1 NP_001106985.1 6 - 7 768 102
NM_001320851.1 NP_001307780.1 5 - 6 549 102
NM_001320852.1 NP_001307781.1 7 - 8 1763 102
NM_001320853.1 NP_001307782.1 6 - 7 674 102
NM_001320854.1 NP_001307783.1 2 - 3 288 102
NM_003899.3 NP_003890.1 6 - 7 881 102
NM_145735.3 NP_663788.1 7 - 8 1763 102
XM_006719956.3 XP_006720019.1 7 - 8 891 102
XM_011521133.2 XP_011519435.1 7 - 8 891 102
XM_017020814.1 XP_016876303.1 7 - 8 891 102
XM_017020815.1 XP_016876304.1 5 - 6 884 102
XM_017020816.1 XP_016876305.1 6 - 7 659 102
XM_017020817.1 XP_016876306.1 5 - 6 884 102
XM_017020818.1 XP_016876307.1 7 - 8 759 102
XM_017020819.1 XP_016876308.1 6 - 7 768 102
XM_017020820.1 XP_016876309.1 6 - 7 891 102
XM_017020821.1 XP_016876310.1 6 - 7 586 102
XM_017020822.1 XP_016876311.1 5 - 6 582 102
XM_017020823.1 XP_016876312.1 5 - 6 1359 102
XM_017020824.1 XP_016876313.1 7 - 8 759 102
XM_017020825.1 XP_016876314.1 5 - 6 510 102
XM_017020826.1 XP_016876315.1 7 - 8 759 102
XM_017020827.1 XP_016876316.1 5 - 6 583 102
XM_017020828.1 XP_016876317.1 2 - 3 148 102
XM_017020829.1 XP_016876318.1 2 - 3 148 102
GenBank mRNA AB177848.1 - 7 - 8 1764 102
AB177849.1 - 4 - 5 487 102
AK295472.1 - 2 - 3 213 102
AK300272.1 - 6 - 7 661 102
AK308897.1 - 2 - 3 288 102
AL834228.1 - 7 - 8 947 102
BC033905.1 - 6 - 7 873 102
BC050521.1 - 6 - 7 740 102
BC060776.1 - 8 - 9 1168 102
D63476.1 - 6 - 7 857 102

View Full Product Details