Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153457026-153457516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS7
Entrez Gene ID
55559
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
UCHL5IP, UIP1
UniGene
Hs.170835
Gene Chromosome Location
Chr.X: 153447665-153495525 on Build GRCh38
Transcripts
NM_017518.7 NR_073156.1

SNP Detail

SNP ID Chromosome Location
rs1051558469 Chr.X: 153457295-153457295 on Build GRCh38
rs946225729 Chr.X: 153457100-153457100 on Build GRCh38
rs781992743 Chr.X: 153457232-153457232 on Build GRCh38
rs981583246 Chr.X: 153457456-153457456 on Build GRCh38
rs73640039 Chr.X: 153457051-153457051 on Build GRCh38
rs782554478 Chr.X: 153457438-153457438 on Build GRCh38
rs1031979273 Chr.X: 153457049-153457049 on Build GRCh38
rs377562227 Chr.X: 153457101-153457101 on Build GRCh38
rs111918191 Chr.X: 153457217-153457217 on Build GRCh38
rs781917859 Chr.X: 153457195-153457195 on Build GRCh38
rs782549606 Chr.X: 153457384-153457384 on Build GRCh38
rs368570184 Chr.X: 153457051-153457051 on Build GRCh38
rs781877970 Chr.X: 153457119-153457119 on Build GRCh38
rs1020529850 Chr.X: 153457448-153457448 on Build GRCh38
rs781928629 Chr.X: 153457086-153457086 on Build GRCh38
rs897687781 Chr.X: 153457274-153457274 on Build GRCh38
rs373741337 Chr.X: 153457220-153457220 on Build GRCh38
rs782155137 Chr.X: 153457139-153457139 on Build GRCh38
rs782776857 Chr.X: 153457303-153457303 on Build GRCh38
rs782450818 Chr.X: 153457158-153457158 on Build GRCh38
rs888877461 Chr.X: 153457297-153457297 on Build GRCh38
rs199939361 Chr.X: 153457140-153457140 on Build GRCh38
rs185205846 Chr.X: 153457275-153457275 on Build GRCh38
rs782650640 Chr.X: 153457194-153457194 on Build GRCh38
rs142983892 Chr.X: 153457226-153457226 on Build GRCh38
rs781796940 Chr.X: 153457143-153457143 on Build GRCh38
rs781806801 Chr.X: 153457398-153457398 on Build GRCh38
rs146365634 Chr.X: 153457216-153457216 on Build GRCh38
rs781836907 Chr.X: 153457166-153457166 on Build GRCh38
rs957712101 Chr.X: 153457420-153457420 on Build GRCh38
rs17091333 Chr.X: 153457452-153457452 on Build GRCh38
rs1012273552 Chr.X: 153457433-153457433 on Build GRCh38
rs146151220 Chr.X: 153457387-153457387 on Build GRCh38
rs782513065 Chr.X: 153457190-153457190 on Build GRCh38
rs111266201 Chr.X: 153457381-153457381 on Build GRCh38
rs928777539 Chr.X: 153457469-153457469 on Build GRCh38
rs782611407 Chr.X: 153457165-153457165 on Build GRCh38
rs111366723 Chr.X: 153457488-153457488 on Build GRCh38
rs782768853 Chr.X: 153457103-153457103 on Build GRCh38
rs990396868 Chr.X: 153457056-153457056 on Build GRCh38
rs782361737 Chr.X: 153457045-153457046 on Build GRCh38
rs913417610 Chr.X: 153457059-153457059 on Build GRCh38
rs375635764 Chr.X: 153457272-153457272 on Build GRCh38
rs782263587 Chr.X: 153457252-153457252 on Build GRCh38
rs782139789 Chr.X: 153457082-153457082 on Build GRCh38

View Full Product Details