The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199563.1 | NP_001186492.1 | 3 - 4 | 569 | 80 |
NM_007073.4 | NP_009004.2 | 3 - 4 | 439 | 80 | |
NM_147147.3 | NP_671488.1 | 3 - 4 | 509 | 80 | |
XM_011535398.2 | XP_011533700.1 | 3 - 4 | 444 | 80 | |
GenBank mRNA | AK289979.1 | - | 3 - 4 | 553 | 80 |
BC034425.1 | - | 3 - 4 | 509 | 80 | |
BC040502.1 | - | 3 - 4 | 569 | 80 |