The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014611.2 | NP_055426.1 | 100 - 101 | 16650 | 64 |
XM_005248700.3 | XP_005248757.1 | 61 - 62 | 10582 | 64 | |
XM_006715405.3 | XP_006715468.1 | 64 - 65 | 12128 | 64 | |
XM_011535635.2 | XP_011533937.1 | 100 - 101 | 16455 | 64 | |
GenBank mRNA | AB002299.2 | - | 54 - 55 | 9306 | 64 |
AF503925.1 | - | 100 - 101 | 16579 | 64 | |
AK027048.1 | - | 4 - 5 | 390 | 64 | |
BC020432.1 | - | 6 - 7 | 633 | 64 | |
BC036888.1 | - | 5 - 6 | 490 | 64 | |
CR627399.1 | - | 46 - 47 | 8105 | 64 |