Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231364426-231364946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGLN1
Entrez Gene ID
54583
Gene Name
egl-9 family hypoxia inducible factor 1
Gene Aliases
C1orf12, ECYT3, HALAH, HIF-PH2, HIFPH2, HPH-2, HPH2, PHD2, SM20, ZMYND6
UniGene
Hs.444450
Gene Chromosome Location
Chr.1: 231363751-231425044 on Build GRCh38
Transcripts
NM_022051.2 XM_005273166.4 XM_005273167.4

SNP Detail

SNP ID Chromosome Location
rs1027737946 Chr.1: 231364672-231364672 on Build GRCh38
rs994572888 Chr.1: 231364648-231364650 on Build GRCh38
rs1045900864 Chr.1: 231364819-231364819 on Build GRCh38
rs979505277 Chr.1: 231364815-231364815 on Build GRCh38
rs968538376 Chr.1: 231364800-231364800 on Build GRCh38
rs41303095 Chr.1: 231364492-231364492 on Build GRCh38
rs748209019 Chr.1: 231364757-231364757 on Build GRCh38
rs774937283 Chr.1: 231364557-231364557 on Build GRCh38
rs913972035 Chr.1: 231364507-231364513 on Build GRCh38
rs895690491 Chr.1: 231364580-231364580 on Build GRCh38
rs917159079 Chr.1: 231364873-231364873 on Build GRCh38
rs139634340 Chr.1: 231364521-231364521 on Build GRCh38
rs1014466968 Chr.1: 231364746-231364746 on Build GRCh38
rs560290409 Chr.1: 231364542-231364542 on Build GRCh38
rs3215625 Chr.1: 231364614-231364615 on Build GRCh38
rs1019154867 Chr.1: 231364738-231364738 on Build GRCh38
rs949919929 Chr.1: 231364918-231364921 on Build GRCh38
rs961217844 Chr.1: 231364604-231364604 on Build GRCh38
rs551676309 Chr.1: 231364702-231364702 on Build GRCh38
rs182146084 Chr.1: 231364803-231364803 on Build GRCh38
rs952105549 Chr.1: 231364914-231364914 on Build GRCh38
rs1001601440 Chr.1: 231364831-231364831 on Build GRCh38
rs1024436657 Chr.1: 231364476-231364476 on Build GRCh38
rs908453718 Chr.1: 231364571-231364571 on Build GRCh38
rs190617118 Chr.1: 231364631-231364631 on Build GRCh38
rs531484702 Chr.1: 231364697-231364697 on Build GRCh38
rs1053391886 Chr.1: 231364709-231364709 on Build GRCh38
rs562736932 Chr.1: 231364662-231364662 on Build GRCh38
rs1036216188 Chr.1: 231364923-231364923 on Build GRCh38
rs1049880297 Chr.1: 231364660-231364660 on Build GRCh38
rs1028080565 Chr.1: 231364834-231364834 on Build GRCh38
rs13239 Chr.1: 231364531-231364531 on Build GRCh38
rs1021053732 Chr.1: 231364789-231364789 on Build GRCh38
rs991427910 Chr.1: 231364851-231364851 on Build GRCh38
rs932380981 Chr.1: 231364659-231364659 on Build GRCh38
rs1001947415 Chr.1: 231364500-231364500 on Build GRCh38
rs533165406 Chr.1: 231364827-231364827 on Build GRCh38
rs755194746 Chr.1: 231364627-231364627 on Build GRCh38
rs542483126 Chr.1: 231364820-231364820 on Build GRCh38
rs553402212 Chr.1: 231364828-231364828 on Build GRCh38
rs758564357 Chr.1: 231364919-231364919 on Build GRCh38
rs1007800761 Chr.1: 231364717-231364717 on Build GRCh38
rs759117979 Chr.1: 231364680-231364680 on Build GRCh38
rs143135847 Chr.1: 231364624-231364624 on Build GRCh38
rs368824788 Chr.1: 231364560-231364560 on Build GRCh38
rs541569859 Chr.1: 231364607-231364608 on Build GRCh38
rs886046097 Chr.1: 231364500-231364500 on Build GRCh38
rs767096160 Chr.1: 231364787-231364787 on Build GRCh38
rs368862498 Chr.1: 231364894-231364894 on Build GRCh38
rs988279253 Chr.1: 231364764-231364764 on Build GRCh38
rs892053026 Chr.1: 231364720-231364720 on Build GRCh38
rs988225965 Chr.1: 231364498-231364498 on Build GRCh38

View Full Product Details