Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231365786-231366287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGLN1
Entrez Gene ID
54583
Gene Name
egl-9 family hypoxia inducible factor 1
Gene Aliases
C1orf12, ECYT3, HALAH, HIF-PH2, HIFPH2, HPH-2, HPH2, PHD2, SM20, ZMYND6
UniGene
Hs.444450
Gene Chromosome Location
Chr.1: 231363751-231425044 on Build GRCh38
Transcripts
NM_022051.2 XM_005273166.4 XM_005273167.4

SNP Detail

SNP ID Chromosome Location
rs1055315500 Chr.1: 231365894-231365894 on Build GRCh38
rs886046107 Chr.1: 231365990-231365990 on Build GRCh38
rs560150547 Chr.1: 231365995-231365995 on Build GRCh38
rs1017422269 Chr.1: 231366000-231366004 on Build GRCh38
rs192117490 Chr.1: 231366024-231366024 on Build GRCh38
rs974916553 Chr.1: 231365977-231365977 on Build GRCh38
rs757801410 Chr.1: 231366121-231366121 on Build GRCh38
rs886046106 Chr.1: 231365876-231365876 on Build GRCh38
rs1055770939 Chr.1: 231366206-231366206 on Build GRCh38
rs1025044054 Chr.1: 231365931-231365931 on Build GRCh38
rs886046108 Chr.1: 231366148-231366150 on Build GRCh38
rs755882450 Chr.1: 231365823-231365823 on Build GRCh38
rs144067360 Chr.1: 231365917-231365917 on Build GRCh38
rs373302655 Chr.1: 231365956-231365957 on Build GRCh38
rs1027364982 Chr.1: 231365963-231365963 on Build GRCh38
rs895286463 Chr.1: 231365914-231365914 on Build GRCh38
rs569310146 Chr.1: 231366095-231366095 on Build GRCh38
rs886046105 Chr.1: 231365852-231365856 on Build GRCh38
rs373743992 Chr.1: 231365836-231365836 on Build GRCh38
rs549470454 Chr.1: 231366031-231366031 on Build GRCh38
rs964497999 Chr.1: 231366003-231366003 on Build GRCh38
rs552048121 Chr.1: 231366180-231366180 on Build GRCh38
rs529087573 Chr.1: 231365916-231365916 on Build GRCh38
rs747131134 Chr.1: 231366058-231366058 on Build GRCh38
rs370227263 Chr.1: 231365814-231365814 on Build GRCh38
rs547609538 Chr.1: 231365942-231365942 on Build GRCh38
rs183940450 Chr.1: 231366099-231366099 on Build GRCh38
rs564839436 Chr.1: 231365887-231365887 on Build GRCh38
rs923016967 Chr.1: 231366040-231366040 on Build GRCh38
rs936951875 Chr.1: 231366155-231366155 on Build GRCh38
rs977788797 Chr.1: 231366077-231366077 on Build GRCh38
rs533283653 Chr.1: 231366062-231366062 on Build GRCh38
rs781574571 Chr.1: 231366238-231366238 on Build GRCh38
rs561398603 Chr.1: 231365883-231365883 on Build GRCh38
rs895338716 Chr.1: 231366218-231366218 on Build GRCh38
rs186972462 Chr.1: 231365959-231365959 on Build GRCh38
rs952017547 Chr.1: 231365985-231365985 on Build GRCh38
rs755084960 Chr.1: 231365848-231365848 on Build GRCh38

View Full Product Details