Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 46550464-46549938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN2
Entrez Gene ID
9721
Gene Name
G protein regulated inducer of neurite outgrowth 2
Gene Aliases
GRIN2, KIAA0514
UniGene
Hs.523375
Gene Chromosome Location
Chr.10: 46543963-46556637 on Build GRCh38
Transcripts
NM_014696.3 XM_005277666.3 XM_011540388.2 XM_011540395.2 XM_011540400.2 XM_017016974.1 XM_017016975.1 XM_017016976.1 XM_017016977.1 XM_017016978.1 XM_017016979.1 XM_017016980.1 XM_017016981.1 XM_017016982.1 XM_017016983.1 XM_017016984.1 XM_017017029.1 XM_017017030.1 XM_017017031.1 XM_017017032.1 XM_017017033.1 XM_017017034.1 XM_017017038.1 XM_017017039.1 XM_017017040.1

Gene Symbol
LOC107984026
Entrez Gene ID
107984026
Gene Name
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.10: 46335552-46838268 on Build GRCh38
Transcripts
NM_014696.3 XM_005277666.3 XM_011540388.2 XM_011540395.2 XM_011540400.2 XM_017016974.1 XM_017016975.1 XM_017016976.1 XM_017016977.1 XM_017016978.1 XM_017016979.1 XM_017016980.1 XM_017016981.1 XM_017016982.1 XM_017016983.1 XM_017016984.1 XM_017017029.1 XM_017017030.1 XM_017017031.1 XM_017017032.1 XM_017017033.1 XM_017017034.1 XM_017017038.1 XM_017017039.1 XM_017017040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355149 c.584C>T p.S195L Chr.10: 46550153-46550153 on Build GRCh38
4144705 c.310A>G p.S104G Chr.10: 46550427-46550427 on Build GRCh38
7420794 c.610A>T p.T204S Chr.10: 46550127-46550127 on Build GRCh38
4933368 c.577C>G p.Q193E Chr.10: 46550160-46550160 on Build GRCh38
6303995 c.722T>C p.V241A Chr.10: 46550015-46550015 on Build GRCh38
View More

View Full Product Details