Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 10 - 11 3056 67
XM_005273164.2 XP_005273221.1 10 - 11 3120 67
XM_005273165.3 XP_005273222.1 10 - 11 3120 67
GenBank mRNA AB007932.1 - 10 - 11 2507 67
BC132676.1 - 10 - 11 2411 67
BC136530.1 - 10 - 11 2411 67
X87831.2 - 8 - 9 961 67

View Full Product Details