Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 11 - 12 3157 110
XM_005273164.2 XP_005273221.1 11 - 12 3221 110
XM_005273165.3 XP_005273222.1 11 - 12 3221 110
GenBank mRNA AB007932.1 - 11 - 12 2608 110
BC132676.1 - 11 - 12 2512 110
BC136530.1 - 11 - 12 2512 110
X87831.2 - 9 - 10 1062 110

View Full Product Details