Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 26 - 27 5522 83
XM_005273164.2 XP_005273221.1 26 - 27 5586 83
XM_005273165.3 XP_005273222.1 26 - 27 5586 83
GenBank mRNA AB007932.1 - 26 - 27 4973 83
BC006193.2 - 4 - 5 437 83
BC009343.2 - 9 - 10 1512 83
BC132676.1 - 26 - 27 4877 83
BC136530.1 - 26 - 27 4877 83
X87831.2 - 24 - 25 3427 83

View Full Product Details