The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111125.2 | NP_001104595.1 | 1 - 2 | 911 | 88 |
GenBank mRNA | AB011094.1 | - | 1 - 2 | 911 | 88 |