Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PHLDB1
- Entrez Gene ID
-
23187
- Gene Name
- pleckstrin homology like domain family B member 1
- Gene Aliases
-
LL5A
- Chromosome Location
-
Chr.11: 118606296 - 118658038 on Build GRCh38
View Full Product Details