The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130965.2 | NP_001124437.1 | 18 - 19 | 2260 | 121 |
NM_001171944.1 | NP_001165415.1 | 16 - 17 | 2010 | 121 | |
NM_025154.5 | NP_079430.3 | 17 - 18 | 2123 | 121 | |
GenBank mRNA | AB018353.1 | - | 19 - 20 | 2358 | 121 |
AB648918.1 | - | 20 - 21 | 2549 | 121 | |
AF202724.1 | - | 9 - 10 | 1159 | 121 | |
AK022816.1 | - | 17 - 18 | 2115 | 121 | |
AK025737.1 | - | 15 - 16 | 1785 | 121 | |
AK074320.1 | - | 5 - 6 | 707 | 121 | |
AK128170.1 | - | 2 - 3 | 1745 | 121 | |
AK302896.1 | - | 16 - 17 | 1982 | 121 | |
AK304150.1 | - | 14 - 15 | 1651 | 121 | |
AY776160.1 | - | 3 - 4 | 348 | 121 | |
BC142707.1 | - | 18 - 19 | 2270 | 121 | |
BC152418.1 | - | 18 - 19 | 2358 | 121 | |
BX640866.1 | - | 9 - 10 | 2349 | 121 | |
BX648893.1 | - | 9 - 10 | 2349 | 121 | |
BX649069.1 | - | 16 - 17 | 2015 | 121 |