The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005256563.4 | XP_005256620.1 | 16 - 17 | 6366 | 83 |
XM_011523761.2 | XP_011522063.1 | 16 - 17 | 6737 | 83 | |
XM_011523762.2 | XP_011522064.1 | 16 - 17 | 6720 | 83 | |
XM_011523763.2 | XP_011522065.1 | 15 - 16 | 6606 | 83 | |
XM_011523764.2 | XP_011522066.1 | 16 - 17 | 6720 | 83 | |
XM_011523765.2 | XP_011522067.1 | 16 - 17 | 6720 | 83 | |
GenBank mRNA | AB020671.2 | - | 1 - 2 | 3258 | 83 |