The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161661.1 | NP_001155133.1 | 22 - 23 | 3382 | 74 |
NM_001161662.1 | NP_001155134.1 | 22 - 23 | 3379 | 74 | |
NM_015238.2 | NP_056053.1 | 22 - 23 | 3364 | 74 | |
XM_005265850.1 | XP_005265907.1 | 22 - 23 | 3354 | 74 | |
XM_005265853.2 | XP_005265910.1 | 22 - 23 | 3318 | 74 | |
XM_011534485.1 | XP_011532787.1 | 24 - 25 | 3537 | 74 | |
XM_011534486.1 | XP_011532788.1 | 24 - 25 | 3534 | 74 | |
XM_011534487.1 | XP_011532789.1 | 24 - 25 | 3522 | 74 | |
XM_011534488.1 | XP_011532790.1 | 24 - 25 | 3519 | 74 | |
XM_011534489.1 | XP_011532791.1 | 24 - 25 | 3519 | 74 | |
XM_011534490.1 | XP_011532792.1 | 24 - 25 | 3516 | 74 | |
XM_011534491.1 | XP_011532793.1 | 24 - 25 | 3504 | 74 | |
XM_017009276.1 | XP_016864765.1 | 24 - 25 | 3501 | 74 | |
XM_017009277.1 | XP_016864766.1 | 24 - 25 | 3900 | 74 | |
XM_017009278.1 | XP_016864767.1 | 22 - 23 | 3789 | 74 | |
GenBank mRNA | AB020676.1 | - | 17 - 18 | 2597 | 74 |
AF506799.1 | - | 22 - 23 | 3337 | 74 | |
AF530058.1 | - | 17 - 18 | 2668 | 74 | |
AK001727.1 | - | 16 - 17 | 2096 | 74 | |
AK027022.1 | - | 9 - 10 | 1263 | 74 | |
AK126376.1 | - | 21 - 22 | 3111 | 74 | |
AK296323.1 | - | 22 - 23 | 3292 | 74 | |
BC004394.1 | - | 7 - 8 | 929 | 74 | |
BC017746.1 | - | 6 - 7 | 789 | 74 | |
BC038463.1 | - | 5 - 6 | 702 | 74 | |
BX640827.1 | - | 22 - 23 | 3183 | 74 |