Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
Chromosome Location
Chr.22: 45192242 - 45240769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009880.1 NP_001009880.1 2 - 3 286 78
NM_015264.1 NP_056079.1 2 - 3 358 78
XM_011530017.2 XP_011528319.1 2 - 3 321 78
XM_017028672.1 XP_016884161.1 2 - 3 330 78
GenBank mRNA AB023147.1 - 1 - 2 54 78
AI830875.1 - 2 - 3 354 78
AK094937.1 - 3 - 4 369 78
AK124648.1 - 2 - 3 366 78
AK297058.1 - 2 - 3 338 78
AK309015.1 - 1 - 2 410 78
AK315883.1 - 2 - 3 270 78
BC038414.1 - 1 - 2 433 78
BC053595.1 - 2 - 3 280 78
CR456402.1 - 2 - 3 223 78
KC154032.1 - 2 - 3 286 78

View Full Product Details