The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009880.1 | NP_001009880.1 | 6 - 7 | 726 | 81 |
NM_015264.1 | NP_056079.1 | 6 - 7 | 798 | 81 | |
XM_011530017.2 | XP_011528319.1 | 6 - 7 | 761 | 81 | |
XM_017028672.1 | XP_016884161.1 | 6 - 7 | 770 | 81 | |
GenBank mRNA | AB023147.1 | - | 5 - 6 | 494 | 81 |
AK025608.1 | - | 4 - 5 | 315 | 81 | |
AK094937.1 | - | 7 - 8 | 809 | 81 | |
AK124648.1 | - | 6 - 7 | 806 | 81 | |
AK297058.1 | - | 6 - 7 | 778 | 81 | |
AK309015.1 | - | 5 - 6 | 850 | 81 | |
AK315883.1 | - | 6 - 7 | 710 | 81 | |
BC016882.1 | - | 1 - 2 | 56 | 81 | |
BC038414.1 | - | 5 - 6 | 873 | 81 | |
BC053595.1 | - | 6 - 7 | 720 | 81 | |
CR456402.1 | - | 6 - 7 | 663 | 81 | |
KC154032.1 | - | 6 - 7 | 726 | 81 |