The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206999.1 | NP_001193928.1 | 46 - 47 | 5939 | 68 |
NM_007174.2 | NP_009105.1 | 45 - 46 | 5813 | 68 | |
XM_006719206.2 | XP_006719269.1 | 46 - 47 | 5921 | 68 | |
XM_011537783.1 | XP_011536085.1 | 47 - 48 | 6011 | 68 | |
XM_011537785.1 | XP_011536087.1 | 47 - 48 | 5966 | 68 | |
XM_011537787.1 | XP_011536089.1 | 45 - 46 | 5795 | 68 | |
XM_011537789.1 | XP_011536091.1 | 39 - 40 | 5146 | 68 | |
XM_011537790.2 | XP_011536092.1 | 38 - 39 | 4714 | 68 | |
XM_011537791.2 | XP_011536093.1 | 38 - 39 | 5699 | 68 | |
XM_017018738.1 | XP_016874227.1 | 36 - 37 | 5255 | 68 | |
GenBank mRNA | AY257469.1 | - | 45 - 46 | 5810 | 68 |
AY681966.1 | - | 46 - 47 | 5939 | 68 |