The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012306.3 | NP_036438.2 | 2 - 3 | 401 | 98 |
XM_017019040.1 | XP_016874529.1 | 3 - 4 | 508 | 98 | |
GenBank mRNA | AB023167.1 | - | 2 - 3 | 298 | 98 |
AF190461.1 | - | 2 - 3 | 307 | 97 | |
AK090728.1 | - | 1 - 2 | 1255 | 98 | |
AK290031.1 | - | 2 - 3 | 304 | 98 | |
AL707764.1 | - | 2 - 3 | 365 | 98 | |
BC000051.2 | - | 2 - 3 | 299 | 98 |