Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYT1L
- Entrez Gene ID
-
23040
- Gene Name
- myelin transcription factor 1 like
- Gene Aliases
-
MRD39, NZF1, ZC2H2C2, ZC2HC4B
- Chromosome Location
-
Chr.2: 1789113 - 2331388 on Build GRCh38
View Full Product Details