Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 10 - 11 1507 70
XM_005267064.3 XP_005267121.1 9 - 10 3586 70
XM_005267065.3 XP_005267122.1 11 - 12 1533 70
XM_005267066.2 XP_005267123.1 9 - 10 2390 70
XM_006715521.1 XP_006715584.1 11 - 12 1760 70
XM_006715522.3 XP_006715585.1 9 - 10 3627 70
XM_011535981.2 XP_011534283.1 9 - 10 4383 70
XM_011535982.2 XP_011534284.1 9 - 10 4383 70
XM_011535983.2 XP_011534285.1 10 - 11 2808 70
XM_017011102.1 XP_016866591.1 9 - 10 2392 70
GenBank mRNA AB033035.1 - 3 - 4 380 70
AJ420468.1 - 7 - 8 745 70
AK056300.1 - 9 - 10 1406 70
AK293288.1 - 9 - 10 1438 70
AK294798.1 - 9 - 10 1561 70
AK307631.1 - 9 - 10 1460 70
BC089428.1 - 5 - 6 618 70
BC140864.1 - 8 - 9 1244 70
BC151134.1 - 8 - 9 1244 70

View Full Product Details