The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015656.1 | NP_056471.1 | 10 - 11 | 2031 | 57 |
XM_011536641.2 | XP_011534943.1 | 7 - 8 | 1176 | 57 | |
GenBank mRNA | AB033062.2 | - | 9 - 10 | 1905 | 57 |
AB290161.1 | - | 9 - 10 | 2001 | 57 |