Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65774315-65774858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5B1
Entrez Gene ID
91056
Gene Name
adaptor related protein complex 5 beta 1 subunit
Gene Aliases
AP-5, PP1030
UniGene
Hs.591957
Gene Chromosome Location
Chr.11: 65773898-65780591 on Build GRCh38
Transcripts
NM_138368.4

SNP Detail

SNP ID Chromosome Location
rs1022272612 Chr.11: 65774346-65774346 on Build GRCh38
rs554766011 Chr.11: 65774615-65774615 on Build GRCh38
rs527242127 Chr.11: 65774512-65774512 on Build GRCh38
rs902339933 Chr.11: 65774413-65774419 on Build GRCh38
rs752164364 Chr.11: 65774699-65774699 on Build GRCh38
rs998277619 Chr.11: 65774710-65774710 on Build GRCh38
rs1012897664 Chr.11: 65774449-65774449 on Build GRCh38
rs916011032 Chr.11: 65774434-65774434 on Build GRCh38
rs567621183 Chr.11: 65774438-65774438 on Build GRCh38
rs967723111 Chr.11: 65774808-65774808 on Build GRCh38
rs919782803 Chr.11: 65774457-65774457 on Build GRCh38
rs151204562 Chr.11: 65774598-65774598 on Build GRCh38
rs943680948 Chr.11: 65774661-65774661 on Build GRCh38
rs148656265 Chr.11: 65774571-65774571 on Build GRCh38
rs985503949 Chr.11: 65774560-65774560 on Build GRCh38
rs866298251 Chr.11: 65774350-65774350 on Build GRCh38
rs973471259 Chr.11: 65774801-65774801 on Build GRCh38
rs1027746594 Chr.11: 65774820-65774820 on Build GRCh38
rs372484582 Chr.11: 65774411-65774411 on Build GRCh38
rs115893008 Chr.11: 65774656-65774656 on Build GRCh38
rs895086689 Chr.11: 65774446-65774446 on Build GRCh38
rs931208813 Chr.11: 65774459-65774459 on Build GRCh38
rs998987439 Chr.11: 65774744-65774744 on Build GRCh38
rs1007903005 Chr.11: 65774508-65774508 on Build GRCh38
rs543787218 Chr.11: 65774686-65774686 on Build GRCh38
rs764738208 Chr.11: 65774599-65774599 on Build GRCh38
rs1014947937 Chr.11: 65774794-65774794 on Build GRCh38
rs547555678 Chr.11: 65774414-65774414 on Build GRCh38
rs1040697782 Chr.11: 65774668-65774668 on Build GRCh38
rs552788922 Chr.11: 65774796-65774796 on Build GRCh38
rs904660902 Chr.11: 65774708-65774708 on Build GRCh38
rs919753297 Chr.11: 65774610-65774610 on Build GRCh38
rs35779020 Chr.11: 65774777-65774778 on Build GRCh38
rs2508301 Chr.11: 65774583-65774583 on Build GRCh38
rs960069938 Chr.11: 65774396-65774396 on Build GRCh38
rs536504494 Chr.11: 65774463-65774463 on Build GRCh38
rs1037099052 Chr.11: 65774700-65774700 on Build GRCh38
rs552017732 Chr.11: 65774341-65774341 on Build GRCh38
rs777967189 Chr.11: 65774687-65774687 on Build GRCh38
rs532792509 Chr.11: 65774705-65774705 on Build GRCh38
rs992861383 Chr.11: 65774580-65774580 on Build GRCh38
rs1021783284 Chr.11: 65774550-65774550 on Build GRCh38
rs886732558 Chr.11: 65774478-65774478 on Build GRCh38
rs908474633 Chr.11: 65774660-65774660 on Build GRCh38
rs968925613 Chr.11: 65774352-65774352 on Build GRCh38
rs924607698 Chr.11: 65774393-65774393 on Build GRCh38
rs916477932 Chr.11: 65774632-65774632 on Build GRCh38
rs563579802 Chr.11: 65774800-65774800 on Build GRCh38
rs779507827 Chr.11: 65774605-65774605 on Build GRCh38
rs181699571 Chr.11: 65774380-65774380 on Build GRCh38
rs199811400 Chr.11: 65774515-65774515 on Build GRCh38
rs763634717 Chr.11: 65774537-65774537 on Build GRCh38
rs932411971 Chr.11: 65774698-65774698 on Build GRCh38
rs946492050 Chr.11: 65774400-65774400 on Build GRCh38
rs569997484 Chr.11: 65774552-65774552 on Build GRCh38
rs35048340 Chr.11: 65774699-65774700 on Build GRCh38
rs494003 Chr.11: 65774827-65774827 on Build GRCh38
rs1031887725 Chr.11: 65774558-65774558 on Build GRCh38
rs1031860075 Chr.11: 65774760-65774760 on Build GRCh38
rs896053244 Chr.11: 65774780-65774780 on Build GRCh38
rs997025444 Chr.11: 65774543-65774543 on Build GRCh38
rs947856704 Chr.11: 65774435-65774435 on Build GRCh38
rs559385450 Chr.11: 65774353-65774353 on Build GRCh38
rs952492873 Chr.11: 65774822-65774822 on Build GRCh38
rs115011816 Chr.11: 65774554-65774554 on Build GRCh38
rs752695908 Chr.11: 65774611-65774611 on Build GRCh38
rs939199107 Chr.11: 65774422-65774422 on Build GRCh38
rs1016133332 Chr.11: 65774518-65774518 on Build GRCh38
rs976918678 Chr.11: 65774542-65774542 on Build GRCh38
rs1044885534 Chr.11: 65774452-65774452 on Build GRCh38
rs550049502 Chr.11: 65774534-65774534 on Build GRCh38

View Full Product Details