Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65777240-65777747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5B1
Entrez Gene ID
91056
Gene Name
adaptor related protein complex 5 beta 1 subunit
Gene Aliases
AP-5, PP1030
UniGene
Hs.591957
Gene Chromosome Location
Chr.11: 65773898-65780591 on Build GRCh38
Transcripts
NM_138368.4

SNP Detail

SNP ID Chromosome Location
rs762416294 Chr.11: 65777468-65777484 on Build GRCh38
rs772601445 Chr.11: 65777375-65777375 on Build GRCh38
rs183505536 Chr.11: 65777674-65777674 on Build GRCh38
rs374297007 Chr.11: 65777361-65777361 on Build GRCh38
rs963209669 Chr.11: 65777480-65777480 on Build GRCh38
rs959428301 Chr.11: 65777474-65777474 on Build GRCh38
rs995819334 Chr.11: 65777442-65777443 on Build GRCh38
rs943146631 Chr.11: 65777385-65777385 on Build GRCh38
rs944845732 Chr.11: 65777638-65777638 on Build GRCh38
rs1043039844 Chr.11: 65777440-65777440 on Build GRCh38
rs943117161 Chr.11: 65777530-65777530 on Build GRCh38
rs139297906 Chr.11: 65777658-65777658 on Build GRCh38
rs1016100100 Chr.11: 65777479-65777479 on Build GRCh38
rs775281874 Chr.11: 65777429-65777429 on Build GRCh38
rs906758247 Chr.11: 65777657-65777657 on Build GRCh38
rs1042619474 Chr.11: 65777633-65777633 on Build GRCh38
rs1009935466 Chr.11: 65777719-65777719 on Build GRCh38
rs963281022 Chr.11: 65777336-65777336 on Build GRCh38
rs555761772 Chr.11: 65777448-65777448 on Build GRCh38
rs559962156 Chr.11: 65777547-65777547 on Build GRCh38
rs949903864 Chr.11: 65777281-65777281 on Build GRCh38
rs919182767 Chr.11: 65777490-65777490 on Build GRCh38
rs537428979 Chr.11: 65777310-65777310 on Build GRCh38
rs937998717 Chr.11: 65777329-65777329 on Build GRCh38
rs1003595022 Chr.11: 65777266-65777266 on Build GRCh38
rs77627301 Chr.11: 65777327-65777327 on Build GRCh38
rs769126554 Chr.11: 65777655-65777655 on Build GRCh38
rs529032160 Chr.11: 65777623-65777623 on Build GRCh38
rs945548208 Chr.11: 65777553-65777553 on Build GRCh38
rs763642177 Chr.11: 65777498-65777498 on Build GRCh38
rs932991071 Chr.11: 65777358-65777358 on Build GRCh38
rs1033308320 Chr.11: 65777690-65777690 on Build GRCh38
rs373036807 Chr.11: 65777636-65777636 on Build GRCh38
rs377444504 Chr.11: 65777400-65777400 on Build GRCh38
rs930682413 Chr.11: 65777620-65777620 on Build GRCh38
rs370026402 Chr.11: 65777541-65777541 on Build GRCh38
rs564625613 Chr.11: 65777481-65777481 on Build GRCh38
rs906505817 Chr.11: 65777451-65777451 on Build GRCh38
rs1047835917 Chr.11: 65777394-65777394 on Build GRCh38
rs899207578 Chr.11: 65777367-65777367 on Build GRCh38
rs972428834 Chr.11: 65777348-65777348 on Build GRCh38
rs575626286 Chr.11: 65777461-65777461 on Build GRCh38
rs563090606 Chr.11: 65777493-65777493 on Build GRCh38
rs1020017524 Chr.11: 65777720-65777720 on Build GRCh38
rs575995291 Chr.11: 65777632-65777632 on Build GRCh38
rs1047783603 Chr.11: 65777631-65777631 on Build GRCh38
rs1034192873 Chr.11: 65777278-65777278 on Build GRCh38
rs920583134 Chr.11: 65777519-65777519 on Build GRCh38
rs1053508773 Chr.11: 65777531-65777531 on Build GRCh38
rs996826350 Chr.11: 65777673-65777673 on Build GRCh38
rs200115064 Chr.11: 65777702-65777702 on Build GRCh38
rs116888179 Chr.11: 65777366-65777366 on Build GRCh38
rs991300241 Chr.11: 65777326-65777326 on Build GRCh38
rs918891775 Chr.11: 65777353-65777353 on Build GRCh38
rs568863133 Chr.11: 65777287-65777287 on Build GRCh38
rs150832077 Chr.11: 65777402-65777402 on Build GRCh38
rs571865787 Chr.11: 65777514-65777514 on Build GRCh38
rs987525050 Chr.11: 65777503-65777503 on Build GRCh38
rs78903837 Chr.11: 65777417-65777417 on Build GRCh38
rs544244726 Chr.11: 65777475-65777475 on Build GRCh38
rs1003971801 Chr.11: 65777457-65777457 on Build GRCh38
rs113195889 Chr.11: 65777319-65777319 on Build GRCh38
rs1019178311 Chr.11: 65777410-65777410 on Build GRCh38
rs769016025 Chr.11: 65777605-65777605 on Build GRCh38

View Full Product Details