The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017554.2 | NP_060024.2 | 13 - 14 | 4263 | 121 |
XM_011512928.2 | XP_011511230.1 | 13 - 14 | 4354 | 121 | |
GenBank mRNA | AB033094.1 | - | 8 - 9 | 2233 | 121 |
AK001770.1 | - | 9 - 10 | 1291 | 121 | |
AK022542.1 | - | 1 - 2 | 68 | 121 | |
AK026003.1 | - | 8 - 9 | 2092 | 121 | |
AK304269.1 | - | 13 - 14 | 4231 | 121 | |
AY134858.1 | - | 8 - 9 | 3436 | 121 | |
BX648758.1 | - | 9 - 10 | 3581 | 121 | |
DQ063584.1 | - | 10 - 11 | 3727 | 121 | |
DQ063585.1 | - | 10 - 11 | 3727 | 121 |