Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
-
Chromosome Location
Chr.4: 87160103 - 87220608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292003.1 NP_001278932.1 9 - 10 2123 94
NM_001292006.1 NP_001278935.1 9 - 10 1852 94
NM_001292007.1 NP_001278936.1 8 - 9 1531 94
NM_020803.4 NP_065854.3 9 - 10 2080 94
NR_120447.1 7 - 8 1344 94
GenBank mRNA AB037799.2 - 8 - 9 1763 94
AK128827.1 - 9 - 10 2123 94
BC041384.2 - 9 - 10 2030 94
BC041901.1 - 7 - 8 1200 94
BX538173.1 - 9 - 10 1997 94
BX640727.1 - 9 - 10 1990 94
BX640744.1 - 9 - 10 1781 94
BX647454.1 - 9 - 10 2015 94
BX648367.1 - 7 - 8 1048 94
CR749265.1 - 8 - 9 1449 94

View Full Product Details