Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67419671-67420173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNS1
Entrez Gene ID
57571
Gene Name
carnosine synthase 1
Gene Aliases
ATPGD1
UniGene
Hs.502982
Gene Chromosome Location
Chr.11: 67415678-67425607 on Build GRCh38
Transcripts
NM_001166222.1 NM_020811.1 XM_011545189.2 XM_011545191.2 XM_017018054.1 XM_017018055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6429694 c.1110C>A p.S370R Chr.11: 67419835-67419835 on Build GRCh38
6315492 c.1069G>T p.A357S Chr.11: 67419794-67419794 on Build GRCh38
4035963 c.1081C>T p.R361W Chr.11: 67419806-67419806 on Build GRCh38
4035962 c.1061G>A p.R354Q Chr.11: 67419786-67419786 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1014039845 Chr.11: 67419845-67419845 on Build GRCh38
rs886639722 Chr.11: 67419886-67419886 on Build GRCh38
rs866149380 Chr.11: 67419830-67419830 on Build GRCh38
rs778084627 Chr.11: 67419694-67419694 on Build GRCh38
rs746358990 Chr.11: 67419774-67419774 on Build GRCh38
rs775201254 Chr.11: 67419734-67419734 on Build GRCh38
rs373121513 Chr.11: 67419841-67419841 on Build GRCh38
rs35876769 Chr.11: 67420115-67420115 on Build GRCh38
rs909849749 Chr.11: 67419719-67419719 on Build GRCh38
rs756073121 Chr.11: 67419749-67419749 on Build GRCh38
rs572926016 Chr.11: 67419728-67419728 on Build GRCh38
rs562912257 Chr.11: 67419951-67419951 on Build GRCh38
rs774161889 Chr.11: 67419798-67419798 on Build GRCh38
rs1051978077 Chr.11: 67419964-67419964 on Build GRCh38
rs747899725 Chr.11: 67419849-67419849 on Build GRCh38
rs751362518 Chr.11: 67419743-67419743 on Build GRCh38
rs201897233 Chr.11: 67419740-67419740 on Build GRCh38
rs145542336 Chr.11: 67419773-67419773 on Build GRCh38
rs12803820 Chr.11: 67419881-67419881 on Build GRCh38
rs922686922 Chr.11: 67419747-67419747 on Build GRCh38
rs752819637 Chr.11: 67419752-67419752 on Build GRCh38
rs199710256 Chr.11: 67419703-67419703 on Build GRCh38
rs761429309 Chr.11: 67419799-67419799 on Build GRCh38
rs527865429 Chr.11: 67420041-67420041 on Build GRCh38
rs767338065 Chr.11: 67419804-67419804 on Build GRCh38
rs750224794 Chr.11: 67419806-67419806 on Build GRCh38
rs1005667151 Chr.11: 67420089-67420089 on Build GRCh38
rs769347606 Chr.11: 67419726-67419726 on Build GRCh38
rs375807563 Chr.11: 67419772-67419772 on Build GRCh38
rs367605521 Chr.11: 67419720-67419720 on Build GRCh38
rs760637952 Chr.11: 67419807-67419807 on Build GRCh38
rs779019452 Chr.11: 67419750-67419750 on Build GRCh38
rs771377503 Chr.11: 67419832-67419832 on Build GRCh38
rs774103222 Chr.11: 67419915-67419915 on Build GRCh38
rs747303499 Chr.11: 67419767-67419767 on Build GRCh38
rs961136632 Chr.11: 67420123-67420123 on Build GRCh38
rs750359189 Chr.11: 67419748-67419748 on Build GRCh38
rs942206733 Chr.11: 67419940-67419940 on Build GRCh38
rs777212446 Chr.11: 67419716-67419716 on Build GRCh38
rs541913714 Chr.11: 67419994-67419994 on Build GRCh38
rs560649410 Chr.11: 67420026-67420026 on Build GRCh38
rs1003427235 Chr.11: 67420040-67420040 on Build GRCh38
rs61891596 Chr.11: 67420090-67420090 on Build GRCh38
rs982045281 Chr.11: 67419894-67419894 on Build GRCh38
rs148879861 Chr.11: 67419955-67419955 on Build GRCh38
rs754816060 Chr.11: 67419731-67419731 on Build GRCh38
rs1052755680 Chr.11: 67420101-67420101 on Build GRCh38
rs954881972 Chr.11: 67420144-67420144 on Build GRCh38
rs371838741 Chr.11: 67419756-67419756 on Build GRCh38
rs766245743 Chr.11: 67419821-67419821 on Build GRCh38
rs781602083 Chr.11: 67419876-67419876 on Build GRCh38
rs906379052 Chr.11: 67420027-67420027 on Build GRCh38
rs760913360 Chr.11: 67419846-67419847 on Build GRCh38
rs200707936 Chr.11: 67419792-67419792 on Build GRCh38
rs564437566 Chr.11: 67420054-67420054 on Build GRCh38
rs1007612294 Chr.11: 67419786-67419786 on Build GRCh38
rs961316120 Chr.11: 67419953-67419954 on Build GRCh38
rs909664822 Chr.11: 67419925-67419925 on Build GRCh38
rs955429333 Chr.11: 67419991-67419991 on Build GRCh38
rs767299540 Chr.11: 67419746-67419746 on Build GRCh38
rs972476722 Chr.11: 67420125-67420125 on Build GRCh38
rs924212707 Chr.11: 67420089-67420089 on Build GRCh38
rs148588805 Chr.11: 67420093-67420093 on Build GRCh38
rs1020874374 Chr.11: 67419847-67419847 on Build GRCh38
rs376841520 Chr.11: 67419704-67419704 on Build GRCh38
rs998512281 Chr.11: 67419820-67419820 on Build GRCh38
rs558877827 Chr.11: 67419741-67419741 on Build GRCh38
rs188621566 Chr.11: 67420048-67420048 on Build GRCh38
rs889086082 Chr.11: 67419982-67419982 on Build GRCh38
rs867998612 Chr.11: 67419903-67419903 on Build GRCh38
rs747395059 Chr.11: 67419695-67419695 on Build GRCh38
rs919396102 Chr.11: 67420126-67420126 on Build GRCh38
rs1023191127 Chr.11: 67419863-67419863 on Build GRCh38
rs561620694 Chr.11: 67420052-67420053 on Build GRCh38
rs777263468 Chr.11: 67419996-67419996 on Build GRCh38
rs757464283 Chr.11: 67419854-67419854 on Build GRCh38
rs771627265 Chr.11: 67419849-67419850 on Build GRCh38
rs761707642 Chr.11: 67419744-67419744 on Build GRCh38
rs79669706 Chr.11: 67419698-67419698 on Build GRCh38
rs1033323631 Chr.11: 67419902-67419902 on Build GRCh38
rs933549493 Chr.11: 67419962-67419962 on Build GRCh38
rs1009389860 Chr.11: 67420015-67420015 on Build GRCh38
rs972235909 Chr.11: 67419941-67419941 on Build GRCh38
rs984292219 Chr.11: 67419735-67419735 on Build GRCh38
rs778559200 Chr.11: 67419818-67419819 on Build GRCh38
rs560398165 Chr.11: 67419995-67419995 on Build GRCh38
rs531920095 Chr.11: 67420096-67420096 on Build GRCh38
rs770256056 Chr.11: 67419782-67419782 on Build GRCh38
rs751778616 Chr.11: 67419853-67419853 on Build GRCh38
rs777999959 Chr.11: 67419764-67419764 on Build GRCh38
rs959067323 Chr.11: 67420071-67420071 on Build GRCh38
rs749115581 Chr.11: 67419785-67419785 on Build GRCh38

View Full Product Details